Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00671228_m1
          See other DAO GE Assays ›
          Gene Symbol:
          Dao
          Gene Name:
          D-amino-acid oxidase
          Gene Aliases:
          Dao1
          Chromosome Location:
          Chr.12: 48353691 - 48373647 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00671228_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dao
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053626.1 NP_446078.1 2 - 3 286 71
          GenBank mRNA AB003400.1 - 2 - 3 286 71
          BC088395.1 - 1 - 2 201 7110523 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200001789.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dao

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          leucine metabolic process
          proline catabolic process
          D-serine catabolic process
          dopamine biosynthetic process
          oxidation-reduction process
          D-alanine catabolic process
          D-serine metabolic process
          D-amino-acid oxidase activity
          receptor binding
          protein dimerization activity
          cofactor binding
          FAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P224631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 309279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132432 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77336 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224631 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P224631 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P224631 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P224631 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701622693 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87201 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84851 - -
          Agilent Rat cDNA Microarray (G4105A) 701622693 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10762885 - -
          Affymetrix Rat Gene 1.0 ST Array 10762884 - -
          Affymetrix Rat Gene 1.0 ST Array 10762886 - -
          Affymetrix Rat Gene 1.0 ST Array 10762873 - -
          Affymetrix Rat Gene 1.0 ST Array 10762874 - -
          Affymetrix Rat Gene 1.0 ST Array 10762883 - -
          Affymetrix Rat Gene 1.0 ST Array 10762875 - -
          Affymetrix Rat Gene 1.0 ST Array 10762879 - -
          Affymetrix Rat Gene 1.0 ST Array 10762872 - -
          Affymetrix Rat Gene 1.0 ST Array 10762882 - -
          Affymetrix Rat Gene 1.0 ST Array 10762878 - -
          Affymetrix Rat Gene 1.0 ST Array 10762877 - -
          Affymetrix Rat Gene 1.0 ST Array 10762881 - -
          Affymetrix Rat Gene 1.0 ST Array 10762876 - -
          Affymetrix Rat Gene 1.0 ST Array 10762880 - -
          Affymetrix Rat Gene 1.1 ST Array 10762871 - -
          Rat Genome 230 2.0 Array 1369491_PM_at - -
          Rat Genome 230 2.0 Array 1369491_at - -
          Affymetrix HT Rat Focus Array 1369491_PM_at - -
          Affymetrix HT Rat Focus Array 1369491_at - -
          Affymetrix Rat Clariom S Assay PSR1200015407.rn.1 TC1200001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015408.rn.1 TC1200001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015410.rn.1 TC1200001789.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200015406.rn.1 TC1200001789.rn.1 RTA TC
          Rat Expression Set 230 1369491_at - -
          Rat Expression Set 230 1369491_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10762871 - -
          Rat Genome U34 Set AB003400_at - -
          Rat Genome U34 Set AB003400_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22339815 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374184 - -
          RatRef-12 Expression BeadChip ILMN_57638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline