Search
Search
| Gene Symbol: Mapk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053842.1 | NP_446294.1 | 6 - 7 | 1025 | 102 | |
| XM_006248658.2 | XP_006248720.1 | 6 - 7 | 1062 | 102 | ||
| XM_006248659.2 | XP_006248721.1 | 6 - 7 | 1062 | 102 | ||
| XM_008768848.1 | XP_008767070.1 | 6 - 7 | 1062 | 102 | ||
| GenBank mRNA | FQ211642.1 | - | 6 - 7 | 1065 | 102 | |
| M64300.1 | - | 6 - 7 | 1025 | 102 | ||
Set Membership: |
|