Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00672436_m1
          See other KCNH6 GE Assays ›
          Gene Symbol:
          Kcnh6
          Gene Name:
          potassium channel, voltage gated eag related subfamily H, member 6
          Gene Aliases:
          Erg2
          Chromosome Location:
          Chr.10: 94207336 - 94228236 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00672436_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcnh6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053937.1 NP_446389.1 5 - 6 1134 75
          XM_006247554.2 XP_006247616.1 5 - 6 1120 75
          XM_006247555.2 XP_006247617.1 5 - 6 1120 75
          XM_006247556.2 XP_006247618.1 5 - 6 1120 75
          XM_008768308.1 XP_008766530.1 5 - 6 1120 75
          XM_008768309.1 XP_008766531.1 5 - 6 1120 75
          GenBank mRNA AF016192.1 - 5 - 6 1134 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000001780.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kcnh6

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          signal transduction
          regulation of ion transmembrane transport
          regulation of membrane potential
          protein heterooligomerization
          potassium ion transmembrane transport
          signal transducer activity
          inward rectifier potassium channel activity
          voltage-gated potassium channel activity
          potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P125350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335210 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84685 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30331 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125350 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P125350 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P125350 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P318308 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701904746 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75634 - -
          Agilent Rat cDNA Microarray (G4105A) 701904746 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739084 - -
          Affymetrix Rat Gene 1.0 ST Array 10739075 - -
          Affymetrix Rat Gene 1.0 ST Array 10739072 - -
          Affymetrix Rat Gene 1.0 ST Array 10739080 - -
          Affymetrix Rat Gene 1.0 ST Array 10739078 - -
          Affymetrix Rat Gene 1.0 ST Array 10739077 - -
          Affymetrix Rat Gene 1.0 ST Array 10739085 - -
          Affymetrix Rat Gene 1.0 ST Array 10739083 - -
          Affymetrix Rat Gene 1.0 ST Array 10739079 - -
          Affymetrix Rat Gene 1.0 ST Array 10739076 - -
          Affymetrix Rat Gene 1.0 ST Array 10739074 - -
          Affymetrix Rat Gene 1.0 ST Array 10739081 - -
          Affymetrix Rat Gene 1.0 ST Array 10739073 - -
          Affymetrix Rat Gene 1.1 ST Array 10739071 - -
          Rat Genome 230 2.0 Array 1369017_PM_at - -
          Rat Genome 230 2.0 Array 1369017_at - -
          Affymetrix HT Rat Focus Array 1369017_at - -
          Affymetrix HT Rat Focus Array 1369017_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000016230.rn.1 TC1000001780.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000016231.rn.1 TC1000001780.rn.1 RTA TC
          Rat Expression Set 230 1369017_PM_at - -
          Rat Expression Set 230 1369017_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739071 - -
          Rat Genome U34 Set rc_AA894087_at - -
          Rat Genome U34 Set rc_AA894087_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373511 - -
          RatRef-12 Expression BeadChip ILMN_58802 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline