Search
Search
| Gene Symbol: Pde10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022236.1 | NP_071572.1 | 20 - 21 | 2462 | 64 | |
| GenBank mRNA | AB027155.1 | - | 20 - 21 | 2462 | 64 | |
| AB027156.1 | - | 2797 | 64 | |||
| AY462091.1 | - | 2428 | 64 | |||
| AY462092.1 | - | 2536 | 64 | |||
| AY462093.1 | - | 2574 | 64 | |||
| AY462094.1 | - | 2243 | 64 | |||
| AY462095.1 | - | 2709 | 64 | |||
| FQ212600.1 | - | 1 - 2 | 69 | 64 | ||
Set Membership: |
|