Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022395.2 | NP_071790.1 | 9 - 10 | 1171 | 66 | |
GenBank mRNA | BC078826.1 | - | 9 - 10 | 1166 | 7113892 | 66 |
D13907.1 | - | 9 - 10 | 1149 | 66 | ||
L12965.1 | - | 9 - 10 | 1172 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |