Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00673737_m1
          See other PLEC GE Assays ›
          Gene Symbol:
          Plec
          Gene Name:
          plectin
          Gene Aliases:
          Pcn, Plec1, Pltn
          Chromosome Location:
          Chr.7: 117230319 - 117291473 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00673737_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plec
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164296.1 NP_001157768.1 19 - 20 2382 77
          NM_001164297.1 NP_001157769.1 19 - 20 2317 77
          NM_001164298.1 NP_001157770.1 19 - 20 2376 77
          NM_001164299.1 NP_001157771.1 19 - 20 2408 77
          NM_001164302.1 NP_001157774.1 19 - 20 2362 77
          NM_001164303.1 NP_001157775.1 19 - 20 2315 77
          NM_001164304.1 NP_001157776.1 19 - 20 2575 77
          NM_001164305.1 NP_001157777.1 19 - 20 2292 77
          NM_001164307.1 NP_001157779.1 19 - 20 2389 77
          NM_001164308.1 NP_001157780.1 19 - 20 2449 77
          NM_022401.2 NP_071796.2 19 - 20 2768 77
          XM_006241870.2 XP_006241932.1 20 - 21 2979 77
          XM_006241875.2 XP_006241937.1 20 - 21 2741 77
          XM_006241882.2 XP_006241944.1 20 - 21 3096 77
          XM_008765592.1 XP_008763814.1 20 - 21 3572 77
          XM_008765593.1 XP_008763815.1 21 - 22 2756 77
          XM_008765594.1 XP_008763816.1 20 - 21 3739 77
          XM_008765595.1 XP_008763817.1 20 - 21 4543 77
          XM_008765596.1 XP_008763818.1 20 - 21 2366 77
          XM_008765597.1 XP_008763819.1 20 - 21 2489 77
          XM_008765598.1 XP_008763820.1 20 - 21 2979 77
          GenBank mRNA AJ635267.2 - 2 - 3 185 77
          AY480022.1 - 19 - 20 2385 77
          AY480023.1 - 19 - 20 2320 77
          AY480024.1 - 19 - 20 2379 77
          AY480025.1 - 19 - 20 2411 77
          AY480026.1 - 19 - 20 2365 77
          AY480027.1 - 19 - 20 2912 77
          AY480028.1 - 19 - 20 2318 77
          AY480029.1 - 19 - 20 2463 77
          AY480030.1 - 19 - 20 2295 77
          AY480031.1 - 19 - 20 2392 77
          AY480032.1 - 19 - 20 2452 77
          X59601.1 - 19 - 20 2718 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003385.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plec

          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          female pregnancy
          response to nutrient
          epithelial cell differentiation
          hemidesmosome assembly
          actin binding
          structural constituent of cytoskeleton
          cytoskeletal protein binding
          structural constituent of muscle
          ankyrin binding
          poly(A) RNA binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044345 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P044351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98390 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58376 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180283 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179952 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31284 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31172 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121496 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044351 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044345 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P044351 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044345 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P044351 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P487852 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31415 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99872 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69002 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72019 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28390 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37808 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904814 - -
          Affymetrix Rat Gene 1.0 ST Array 10904860 - -
          Affymetrix Rat Gene 1.0 ST Array 10904815 - -
          Affymetrix Rat Gene 1.0 ST Array 10904841 - -
          Affymetrix Rat Gene 1.0 ST Array 10904838 - -
          Affymetrix Rat Gene 1.0 ST Array 10904837 - -
          Affymetrix Rat Gene 1.0 ST Array 10904829 - -
          Affymetrix Rat Gene 1.0 ST Array 10904843 - -
          Affymetrix Rat Gene 1.0 ST Array 10904839 - -
          Affymetrix Rat Gene 1.0 ST Array 10904861 - -
          Affymetrix Rat Gene 1.0 ST Array 10904825 - -
          Affymetrix Rat Gene 1.0 ST Array 10904852 - -
          Affymetrix Rat Gene 1.0 ST Array 10904832 - -
          Affymetrix Rat Gene 1.0 ST Array 10904856 - -
          Affymetrix Rat Gene 1.0 ST Array 10904831 - -
          Affymetrix Rat Gene 1.0 ST Array 10904822 - -
          Affymetrix Rat Gene 1.0 ST Array 10904849 - -
          Affymetrix Rat Gene 1.0 ST Array 10904848 - -
          Affymetrix Rat Gene 1.0 ST Array 10904842 - -
          Affymetrix Rat Gene 1.0 ST Array 10904833 - -
          Affymetrix Rat Gene 1.0 ST Array 10904818 - -
          Affymetrix Rat Gene 1.0 ST Array 10904863 - -
          Affymetrix Rat Gene 1.0 ST Array 10904854 - -
          Affymetrix Rat Gene 1.0 ST Array 10904834 - -
          Affymetrix Rat Gene 1.0 ST Array 10904826 - -
          Affymetrix Rat Gene 1.0 ST Array 10904827 - -
          Affymetrix Rat Gene 1.0 ST Array 10904859 - -
          Affymetrix Rat Gene 1.0 ST Array 10904857 - -
          Affymetrix Rat Gene 1.0 ST Array 10904828 - -
          Affymetrix Rat Gene 1.0 ST Array 10904824 - -
          Affymetrix Rat Gene 1.0 ST Array 10904840 - -
          Affymetrix Rat Gene 1.0 ST Array 10904821 - -
          Affymetrix Rat Gene 1.0 ST Array 10904855 - -
          Affymetrix Rat Gene 1.0 ST Array 10904816 - -
          Affymetrix Rat Gene 1.0 ST Array 10904844 - -
          Affymetrix Rat Gene 1.0 ST Array 10904813 - -
          Affymetrix Rat Gene 1.0 ST Array 10904853 - -
          Affymetrix Rat Gene 1.0 ST Array 10904845 - -
          Affymetrix Rat Gene 1.0 ST Array 10904823 - -
          Affymetrix Rat Gene 1.0 ST Array 10904836 - -
          Affymetrix Rat Gene 1.0 ST Array 10904862 - -
          Affymetrix Rat Gene 1.0 ST Array 10904851 - -
          Affymetrix Rat Gene 1.0 ST Array 10904835 - -
          Affymetrix Rat Gene 1.0 ST Array 10904846 - -
          Affymetrix Rat Gene 1.0 ST Array 10904847 - -
          Affymetrix Rat Gene 1.0 ST Array 10904819 - -
          Affymetrix Rat Gene 1.0 ST Array 10904850 - -
          Affymetrix Rat Gene 1.0 ST Array 10904817 - -
          Affymetrix Rat Gene 1.0 ST Array 10904858 - -
          Affymetrix Rat Gene 1.0 ST Array 10904820 - -
          Affymetrix Rat Gene 1.0 ST Array 10904830 - -
          Affymetrix Rat Gene 1.1 ST Array 10904812 - -
          Rat Genome 230 2.0 Array 1386941_PM_at - -
          Rat Genome 230 2.0 Array 1386941_at - -
          Affymetrix HT Rat Focus Array 1386941_at - -
          Affymetrix HT Rat Focus Array 1386941_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700026165.rn.1 TC0700003385.rn.1 RTA TC
          Rat Expression Set 230 1386941_PM_at - -
          Rat Expression Set 230 1386941_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904812 - -
          Rat Genome U34 Set X59601_at - -
          Rat Genome U34 Set U96275_at - -
          Rat Genome U34 Set U96275_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21279764 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360481 - -
          RatRef-12 Expression BeadChip ILMN_49489 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline