Search
Search
| Gene Symbol: Ppap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022538.2 | NP_071983.2 | 4 - 5 | 643 | 70 | |
| XM_006231949.2 | XP_006232011.1 | 4 - 5 | 1011 | 70 | ||
| GenBank mRNA | AF503609.1 | - | 4 - 5 | 549 | 70 | |
| AF503610.1 | - | 3 - 4 | 394 | 70 | ||
| BC061815.1 | - | 4 - 5 | 643 | 5598693 | 70 | |
| FQ229687.1 | - | 2 - 3 | 238 | 70 | ||
| JQ013728.1 | - | 4 - 5 | 581 | 70 | ||
| U90556.1 | - | 4 - 5 | 558 | 70 | ||
Set Membership: |
|