Search
Search
| Gene Symbol: Dpysl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023023.1 | NP_075412.1 | 2 - 3 | 389 | 74 | |
| GenBank mRNA | AB029432.1 | - | 1 - 2 | 266 | 74 | |
| AJ131436.1 | - | 2 - 3 | 389 | 74 | ||
Set Membership: |
|