Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00675339_m1
          See other GRPEL1 GE Assays ›
          Gene Symbol:
          Grpel1
          Gene Name:
          GrpE-like 1, mitochondrial
          Gene Aliases:
          GrpE#1, GrpE1
          Chromosome Location:
          Chr.14: 79458950 - 79464775 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00675339_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grpel1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024487.4 NP_077813.1 2 - 3 234 72
          XM_006251142.2 XP_006251204.1 3 - 4 427 72
          GenBank mRNA BC097312.1 - 2 - 3 232 7387647 72
          FQ219742.1 - 3 - 4 415 72
          FQ220295.1 - 2 - 3 225 72
          U62940.1 - 2 - 3 235 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400002054.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grpel1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          regulation of catalytic activity
          adenyl-nucleotide exchange factor activity
          protein homodimerization activity
          unfolded protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P160868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966260 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160868 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P160868 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P160868 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P160868 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6079 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92076 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10777444 - -
          Affymetrix Rat Gene 1.0 ST Array 10777445 - -
          Affymetrix Rat Gene 1.0 ST Array 10777447 - -
          Affymetrix Rat Gene 1.0 ST Array 10777446 - -
          Affymetrix Rat Gene 1.0 ST Array 10777443 - -
          Affymetrix Rat Gene 1.1 ST Array 10777442 - -
          Rat Genome 230 2.0 Array 1368552_at - -
          Rat Genome 230 2.0 Array 1368552_PM_at - -
          Affymetrix HT Rat Focus Array 1368552_PM_at - -
          Affymetrix HT Rat Focus Array 1368552_at - -
          Affymetrix Rat Clariom S Assay PSR1400014594.rn.1 TC1400002054.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014593.rn.1 TC1400002054.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014597.rn.1 TC1400002054.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1400014596.rn.1 TC1400002054.rn.1 RTA TC
          Rat Expression Set 230 1368552_PM_at - -
          Rat Expression Set 230 1368552_at - -
          Affymetrix Rat Gene 1.0 ST Array 10777442 - -
          Rat Genome U34 Set U62940_at - -
          Rat Genome U34 Set rc_AA996644_at - -
          Rat Genome U34 Set U62940_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22036824 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364021 - -
          RatRef-12 Expression BeadChip ILMN_61667 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline