Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00677839_m1
          See other CSNK1A1 GE Assays ›
          Gene Symbol:
          Csnk1a1
          Gene Name:
          casein kinase 1, alpha 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 56887722 - 56910610 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00677839_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Csnk1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053615.1 NP_446067.1 8 - 9 852 79
          GenBank mRNA BC127486.1 - 1078 8364080 79
          U77582.1 - 8 - 9 852 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000630.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Csnk1a1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          protein phosphorylation
          phagocytosis
          mitotic nuclear division
          regulation of cell shape
          Wnt signaling pathway
          peptidyl-serine phosphorylation
          cell division
          magnesium ion binding
          glycoprotein binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          peptide binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P079419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7108 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88918 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079419 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P079419 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P079419 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P164889 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12873 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700529826 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700325233 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15037 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80904 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40654 - -
          Agilent Rat cDNA Microarray (G4105A) 700325233 - -
          Agilent Rat cDNA Microarray (G4105A) 700529826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802118 - -
          Affymetrix Rat Gene 1.0 ST Array 10802119 - -
          Affymetrix Rat Gene 1.0 ST Array 10802115 - -
          Affymetrix Rat Gene 1.0 ST Array 10802121 - -
          Affymetrix Rat Gene 1.0 ST Array 10802117 - -
          Affymetrix Rat Gene 1.0 ST Array 10802123 - -
          Affymetrix Rat Gene 1.0 ST Array 10802122 - -
          Affymetrix Rat Gene 1.0 ST Array 10802127 - -
          Affymetrix Rat Gene 1.0 ST Array 10802116 - -
          Affymetrix Rat Gene 1.0 ST Array 10802124 - -
          Affymetrix Rat Gene 1.0 ST Array 10802125 - -
          Affymetrix Rat Gene 1.0 ST Array 10802126 - -
          Affymetrix Rat Gene 1.1 ST Array 10802114 - -
          Rat Genome 230 2.0 Array 1389859_at - -
          Rat Genome 230 2.0 Array 1381918_at - -
          Rat Genome 230 2.0 Array 1369312_a_at - -
          Rat Genome 230 2.0 Array 1374392_PM_at - -
          Rat Genome 230 2.0 Array 1389859_PM_at - -
          Rat Genome 230 2.0 Array 1374646_at - -
          Rat Genome 230 2.0 Array 1374646_PM_at - -
          Rat Genome 230 2.0 Array 1394818_at - -
          Rat Genome 230 2.0 Array 1369312_PM_a_at - -
          Rat Genome 230 2.0 Array 1394818_PM_at - -
          Rat Genome 230 2.0 Array 1381918_PM_at - -
          Rat Genome 230 2.0 Array 1374392_at - -
          Affymetrix HT Rat Focus Array 1369312_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369312_a_at - -
          Affymetrix HT Rat Focus Array 1374646_PM_at - -
          Affymetrix HT Rat Focus Array 1389859_PM_at - -
          Affymetrix HT Rat Focus Array 1389859_at - -
          Affymetrix HT Rat Focus Array 1381918_at - -
          Affymetrix HT Rat Focus Array 1381918_PM_at - -
          Affymetrix HT Rat Focus Array 1374646_at - -
          Affymetrix Rat Clariom S Assay PSR1800004690.rn.1 TC1800000630.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004697.rn.1 TC1800000630.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800004688.rn.1 TC1800000630.rn.1 RTA TC
          Rat Expression Set 230 1389859_PM_at - -
          Rat Expression Set 230 1369312_PM_a_at - -
          Rat Expression Set 230 1374646_PM_at - -
          Rat Expression Set 230 1369312_a_at - -
          Rat Expression Set 230 1389859_at - -
          Rat Expression Set 230 1374646_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802114 - -
          Rat Genome U34 Set U77583_at - -
          Rat Genome U34 Set rc_AI235291_at - -
          Rat Genome U34 Set rc_AI030738_at - -
          Rat Genome U34 Set rc_AA955819_g_at - -
          Rat Genome U34 Set rc_AA955819_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20802188 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354478 - -
          RatRef-12 Expression BeadChip ILMN_51884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline