Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680558_m1
          See other SCN2A GE Assays ›
          Gene Symbol:
          Scn2a
          Gene Name:
          sodium channel, voltage-gated, type II, alpha subunit
          Gene Aliases:
          NachII, Nav1.2, RII/RIIA, RNSCPIIR, SCN, Scn2a1, Scn2a2, ScpII
          Chromosome Location:
          Chr.3: 51687910 - 51822008 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680558_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012647.1 NP_036779.1 22 - 23 4467 80
          GenBank mRNA M22254.1 - 22 - 23 4466 80
          X03639.1 - 22 - 23 4467 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000706.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn2a

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          nervous system development
          intrinsic apoptotic signaling pathway in response to osmotic stress
          neuronal action potential
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          myelination
          neuron apoptotic process
          membrane depolarization during action potential
          voltage-gated sodium channel activity
          protein binding
          sodium ion binding
          leucine zipper domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P034911 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034911 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P034911 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P034911 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P121187 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P100365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P529319 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836481 - -
          Affymetrix Rat Gene 1.0 ST Array 10836469 - -
          Affymetrix Rat Gene 1.0 ST Array 10836473 - -
          Affymetrix Rat Gene 1.0 ST Array 10836468 - -
          Affymetrix Rat Gene 1.0 ST Array 10836487 - -
          Affymetrix Rat Gene 1.0 ST Array 10836471 - -
          Affymetrix Rat Gene 1.0 ST Array 10836474 - -
          Affymetrix Rat Gene 1.0 ST Array 10836460 - -
          Affymetrix Rat Gene 1.0 ST Array 10836478 - -
          Affymetrix Rat Gene 1.0 ST Array 10836459 - -
          Affymetrix Rat Gene 1.0 ST Array 10836470 - -
          Affymetrix Rat Gene 1.0 ST Array 10836464 - -
          Affymetrix Rat Gene 1.0 ST Array 10836482 - -
          Affymetrix Rat Gene 1.0 ST Array 10836463 - -
          Affymetrix Rat Gene 1.0 ST Array 10836466 - -
          Affymetrix Rat Gene 1.0 ST Array 10836480 - -
          Affymetrix Rat Gene 1.0 ST Array 10836461 - -
          Affymetrix Rat Gene 1.0 ST Array 10836472 - -
          Affymetrix Rat Gene 1.0 ST Array 10836462 - -
          Affymetrix Rat Gene 1.0 ST Array 10836477 - -
          Affymetrix Rat Gene 1.0 ST Array 10836475 - -
          Affymetrix Rat Gene 1.0 ST Array 10836476 - -
          Affymetrix Rat Gene 1.0 ST Array 10836465 - -
          Affymetrix Rat Gene 1.0 ST Array 10836488 - -
          Affymetrix Rat Gene 1.0 ST Array 10836489 - -
          Affymetrix Rat Gene 1.0 ST Array 10836486 - -
          Affymetrix Rat Gene 1.0 ST Array 10836483 - -
          Affymetrix Rat Gene 1.0 ST Array 10836479 - -
          Affymetrix Rat Gene 1.0 ST Array 10836485 - -
          Affymetrix Rat Gene 1.0 ST Array 10836484 - -
          Affymetrix Rat Gene 1.0 ST Array 10836467 - -
          Affymetrix Rat Gene 1.1 ST Array 10836458 - -
          Rat Genome 230 2.0 Array 1369662_PM_at - -
          Rat Genome 230 2.0 Array 1369662_at - -
          Affymetrix HT Rat Focus Array 1369662_PM_at - -
          Affymetrix HT Rat Focus Array 1369662_at - -
          Affymetrix Rat Clariom S Assay PSR0300005333.rn.1 TC0300000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005334.rn.1 TC0300000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005328.rn.1 TC0300000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005335.rn.1 TC0300000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005329.rn.1 TC0300000706.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005332.rn.1 TC0300000706.rn.1 RTA TC
          Rat Expression Set 230 1369662_at - -
          Rat Expression Set 230 1369662_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836458 - -
          Rat Genome U34 Set M22254_at - -
          Rat Genome U34 Set M31433mRNA#1_at - -
          Rat Genome U34 Set M31433mRNA#1_at - -
          Rat Genome U34 Set M22254_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21910933 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352812 - -
          RatRef-12 Expression BeadChip ILMN_57429 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline