Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00680749_g1
          See other BSG GE Assays ›
          Gene Symbol:
          Bsg
          Gene Name:
          basigin (Ok blood group)
          Gene Aliases:
          5A11, CE9, EMMPRIN, HT7, Ox47R, basignin
          Chromosome Location:
          Chr.7: 12875536 - 12882753 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00680749_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bsg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109882.1 NP_001103352.1 7 - 8 1193 74
          NM_012783.3 NP_036915.1 6 - 7 845 74
          GenBank mRNA AY120888.1 - 7 - 8 1112 74
          BC059145.1 - 6 - 7 769 6889785 74
          BQ199464.1 - 1 - 2 74
          FQ212571.1 - 6 - 7 781 74
          FQ212582.1 - 6 - 7 789 74
          FQ213065.1 - 6 - 7 791 74
          FQ214193.1 - 6 - 7 781 74
          FQ214232.1 - 6 - 7 786 74
          FQ218399.1 - 6 - 7 780 74
          FQ219910.1 - 6 - 7 765 74
          FQ220920.1 - 6 - 7 789 74
          FQ224837.1 - 6 - 7 790 74
          FQ229144.1 - 6 - 7 791 74
          FQ229755.1 - 6 - 7 778 74
          FQ231850.1 - 6 - 7 791 74
          FQ232262.1 - 6 - 7 781 74
          FQ234420.1 - 6 - 7 791 74
          FQ234809.1 - 6 - 7 789 74
          X54640.1 - 972 74
          X67215.1 - 6 - 7 766 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700004051.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bsg

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          embryo implantation
          odontogenesis of dentin-containing tooth
          response to peptide hormone
          response to mercury ion
          decidualization
          response to cAMP
          protein targeting to plasma membrane
          protein binding
          mannose binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164788 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013284 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P013284 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P013284 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P498771 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700491717 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702150583 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27511 - -
          Agilent Rat cDNA Microarray (G4105A) 702150583 - -
          Agilent Rat cDNA Microarray (G4105A) 700491717 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900807 - -
          Affymetrix Rat Gene 1.0 ST Array 10900806 - -
          Affymetrix Rat Gene 1.0 ST Array 10900809 - -
          Affymetrix Rat Gene 1.0 ST Array 10900808 - -
          Affymetrix Rat Gene 1.0 ST Array 10900810 - -
          Affymetrix Rat Gene 1.0 ST Array 10900805 - -
          Affymetrix Rat Gene 1.0 ST Array 10900811 - -
          Affymetrix Rat Gene 1.0 ST Array 10900812 - -
          Affymetrix Rat Gene 1.0 ST Array 10900804 - -
          Affymetrix Rat Gene 1.1 ST Array 10900803 - -
          Rat Genome 230 2.0 Array 1367643_PM_at - -
          Rat Genome 230 2.0 Array 1367643_at - -
          Affymetrix HT Rat Focus Array 1367643_PM_at - -
          Affymetrix HT Rat Focus Array 1367643_at - -
          Affymetrix Rat Clariom S Assay PSR0700033257.rn.1 TC0700004051.rn.1 RTA TC
          Rat Expression Set 230 1367643_at - -
          Rat Expression Set 230 1367643_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900803 - -
          Rat Genome U34 Set X54640_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22021861 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357736 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline