Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00681263_g1
          See other DRD4 GE Assays ›
          Gene Symbol:
          Drd4
          Gene Name:
          dopamine receptor D4
          Gene Aliases:
          D4RA
          Chromosome Location:
          Chr.1: 214278296 - 214282818 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00681263_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Drd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012944.1 NP_037076.1 3 - 4 1083 64
          XM_006230521.1 XP_006230583.1 3 - 4 1053 64
          XM_006230522.1 XP_006230584.1 4 - 5 969 64
          GenBank mRNA M84009.1 - 3 - 4 1083 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002893.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Drd4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          behavioral fear response
          synaptic transmission, dopaminergic
          response to amphetamine
          negative regulation of adenylate cyclase activity
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          dopamine receptor signaling pathway
          short-term memory
          circadian rhythm
          locomotory behavior
          associative learning
          adult locomotory behavior
          olfactory learning
          photoperiodism
          negative regulation of cAMP biosynthetic process
          positive regulation of sodium:proton antiporter activity
          positive regulation of kinase activity
          response to histamine
          regulation of dopamine metabolic process
          fear response
          regulation of circadian rhythm
          regulation of neurotransmitter secretion
          behavioral response to cocaine
          response to steroid hormone
          arachidonic acid secretion
          negative regulation of protein secretion
          modulation of synaptic transmission
          regulation of calcium-mediated signaling
          retina development in camera-type eye
          inhibitory postsynaptic potential
          positive regulation of penile erection
          negative regulation of voltage-gated calcium channel activity
          positive regulation of excitatory postsynaptic potential
          dopamine neurotransmitter receptor activity, coupled via Gi/Go
          dopamine neurotransmitter receptor activity
          protein binding
          drug binding
          SH3 domain binding
          dopamine binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P150519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505131 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P150519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P150519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P332729 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16333 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10712269 - -
          Affymetrix Rat Gene 1.0 ST Array 10712267 - -
          Affymetrix Rat Gene 1.0 ST Array 10712266 - -
          Affymetrix Rat Gene 1.0 ST Array 10712268 - -
          Affymetrix Rat Gene 1.1 ST Array 10712265 - -
          Rat Genome 230 2.0 Array 1387520_at - -
          Rat Genome 230 2.0 Array 1394384_PM_at - -
          Rat Genome 230 2.0 Array 1394385_PM_s_at - -
          Rat Genome 230 2.0 Array 1387520_PM_at - -
          Rat Genome 230 2.0 Array 1394385_s_at - -
          Rat Genome 230 2.0 Array 1394384_at - -
          Affymetrix HT Rat Focus Array 1394385_PM_s_at - -
          Affymetrix HT Rat Focus Array 1387520_at - -
          Affymetrix HT Rat Focus Array 1394384_at - -
          Affymetrix HT Rat Focus Array 1394384_PM_at - -
          Affymetrix HT Rat Focus Array 1394385_s_at - -
          Affymetrix HT Rat Focus Array 1387520_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100022952.rn.1 TC0100002893.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022955.rn.1 TC0100002893.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022954.rn.1 TC0100002893.rn.1 RTA TC
          Rat Expression Set 230 1394384_at - -
          Rat Expression Set 230 1394385_PM_s_at - -
          Rat Expression Set 230 1387520_at - -
          Rat Expression Set 230 1387520_PM_at - -
          Rat Expression Set 230 1394385_s_at - -
          Rat Expression Set 230 1394384_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10712265 - -
          Rat Genome U34 Set M84009_at - -
          Rat Genome U34 Set M84009_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22334888 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354010 - -
          RatRef-12 Expression BeadChip ILMN_67874 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline