Search
Search
| Gene Symbol: Dnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013199.1 | NP_037331.1 | 6 - 7 | 962 | 58 | |
| XM_006242604.2 | XP_006242666.1 | 6 - 7 | 1027 | 58 | ||
| XM_006242605.2 | XP_006242667.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242608.2 | XP_006242670.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242610.2 | XP_006242672.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242611.2 | XP_006242673.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242612.2 | XP_006242674.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242613.2 | XP_006242675.1 | 6 - 7 | 1026 | 58 | ||
| XM_006242614.2 | XP_006242676.1 | 6 - 7 | 1026 | 58 | ||
| XM_008765917.1 | XP_008764139.1 | 6 - 7 | 1027 | 58 | ||
| XM_008765918.1 | XP_008764140.1 | 6 - 7 | 1026 | 58 | ||
| XM_008765919.1 | XP_008764141.1 | 6 - 7 | 1026 | 58 | ||
| GenBank mRNA | L24562.1 | - | 6 - 7 | 901 | 58 | |
| L25605.1 | - | 6 - 7 | 962 | 58 | ||
Set Membership: |
|