Search
Search
| Gene Symbol: Akr7a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013215.1 | NP_037347.1 | 5 - 6 | 762 | 74 | |
| GenBank mRNA | AF045464.1 | - | 5 - 6 | 762 | 74 | |
| BC078872.1 | - | 5 - 6 | 756 | 7127297 | 74 | |
| BC089811.1 | - | 5 - 6 | 725 | 7126881 | 74 | |
| X74673.1 | - | 5 - 6 | 759 | 74 | ||
Set Membership: |
|