Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00685720_m1
          See other FAS GE Assays ›
          Gene Symbol:
          Fas
          Gene Name:
          Fas cell surface death receptor
          Gene Aliases:
          Tnfrsf6
          Chromosome Location:
          Chr.1: 252589785 - 252624790 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00685720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fas
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139194.2 NP_631933.2 8 - 9 688 79
          GenBank mRNA D26113.1 - 8 - 9 713 79
          FQ233018.1 - 8 - 9 676 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003512.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fas

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle atresia
          response to hypoxia
          immunoglobulin production
          renal system process
          apoptotic process
          activation-induced cell death of T cells
          inflammatory cell apoptotic process
          obsolete transformed cell apoptotic process
          immune response
          spermatogenesis
          aging
          circadian rhythm
          extrinsic apoptotic signaling pathway via death domain receptors
          response to toxic substance
          response to inorganic substance
          gene expression
          regulation of gene expression
          positive regulation of cell death
          B cell mediated immunity
          telencephalon development
          dendrite regeneration
          positive regulation of protein homooligomerization
          response to lipopolysaccharide
          tumor necrosis factor-mediated signaling pathway
          response to cytokine
          regulation of cell proliferation
          response to drug
          ovulation cycle
          regulation of apoptotic process
          chordate embryonic development
          T cell homeostasis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to peptide hormone
          response to estrogen
          negative thymic T cell selection
          regulation of lymphocyte differentiation
          regulation of myeloid cell differentiation
          response to cycloheximide
          spleen development
          negative regulation of B cell activation
          protein homooligomerization
          response to glucocorticoid
          neuron apoptotic process
          maternal process involved in female pregnancy
          cellular response to hydrogen peroxide
          response to growth factor
          cellular response to phenylalanine
          cellular response to mechanical stimulus
          cellular response to cobalt ion
          cellular response to lithium ion
          cellular response to glucose stimulus
          cellular response to cytokine stimulus
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to estrogen stimulus
          cellular response to organic cyclic compound
          cellular response to hyperoxia
          cellular response to hypoxia
          cellular response to hydrostatic pressure
          motor neuron apoptotic process
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          extrinsic apoptotic signaling pathway in absence of ligand
          hepatocyte apoptotic process
          liver regeneration
          necroptotic signaling pathway
          response to fluoride
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          protease binding
          receptor activity
          transmembrane signaling receptor activity
          tumor necrosis factor-activated receptor activity
          protein binding
          kinase binding
          protein complex binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P463745 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P131736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130304 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156958 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P463745 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P131736 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P463745 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P463745 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P131736 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P652897 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P463745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54654 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1721 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58862 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31887 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10714894 - -
          Affymetrix Rat Gene 1.0 ST Array 10714895 - -
          Affymetrix Rat Gene 1.0 ST Array 10714893 - -
          Affymetrix Rat Gene 1.0 ST Array 10714898 - -
          Affymetrix Rat Gene 1.0 ST Array 10714899 - -
          Affymetrix Rat Gene 1.0 ST Array 10714891 - -
          Affymetrix Rat Gene 1.0 ST Array 10714897 - -
          Affymetrix Rat Gene 1.0 ST Array 10714892 - -
          Affymetrix Rat Gene 1.0 ST Array 10714896 - -
          Affymetrix Rat Gene 1.1 ST Array 10714890 - -
          Rat Genome 230 2.0 Array 1384842_s_at - -
          Rat Genome 230 2.0 Array 1384842_PM_s_at - -
          Rat Genome 230 2.0 Array 1369900_PM_at - -
          Rat Genome 230 2.0 Array 1369900_at - -
          Rat Genome 230 2.0 Array 1397221_s_at - -
          Rat Genome 230 2.0 Array 1397221_PM_s_at - -
          Affymetrix HT Rat Focus Array 1397221_s_at - -
          Affymetrix HT Rat Focus Array 1369900_at - -
          Affymetrix HT Rat Focus Array 1384842_PM_s_at - -
          Affymetrix HT Rat Focus Array 1384842_s_at - -
          Affymetrix HT Rat Focus Array 1369900_PM_at - -
          Affymetrix HT Rat Focus Array 1397221_PM_s_at - -
          Affymetrix Rat Clariom S Assay PSR0100028468.rn.1 TC0100003512.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028467.rn.1 TC0100003512.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100028469.rn.1 TC0100003512.rn.1 RTA TC
          Rat Expression Set 230 1369900_PM_at - -
          Rat Expression Set 230 1369900_at - -
          Rat Expression Set 230 1384842_PM_s_at - -
          Rat Expression Set 230 1397221_PM_s_at - -
          Rat Expression Set 230 1384842_s_at - -
          Rat Expression Set 230 1397221_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10714890 - -
          Rat Genome U34 Set AFFX-MurFAS_at - -
          Rat Genome U34 Set D26112_s_at - -
          Rat Genome U34 Set AFFX-MurFAS_at - -
          Rat Genome U34 Set AFFX-MurFAS_at - -
          Rat Genome U34 Set rc_AI231531_i_at - -
          Rat Genome U34 Set D26112_s_at - -
          Rat Genome U34 Set AFFX-MurFAS_at - -
          Rat Genome U34 Set D26112_s_at - -
          Rat Genome U34 Set AFFX-MurFAS_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21697087 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373348 - -
          RatRef-12 Expression BeadChip ILMN_51642 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline