Search
Search

Macrod1| Gene Symbol: Macrod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139337.1 | NP_647553.1 | 9 - 10 | 770 | 69 | |
| GenBank mRNA | AF404762.1 | - | 9 - 10 | 770 | 69 | |
| AF419856.1 | - | 9 - 10 | 803 | 69 | ||
| FQ216840.1 | - | 9 - 10 | 938 | 69 | ||
Set Membership: |
|