Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00685960_m1
          See other GPSM1 GE Assays ›
          Gene Symbol:
          Gpsm1
          Gene Name:
          G-protein signaling modulator 1
          Gene Aliases:
          Ags3
          Chromosome Location:
          Chr.3: 3767394 - 3794360 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00685960_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpsm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001145469.1 NP_001138941.1 3 - 4 493 124
          NM_144745.2 NP_653346.2 13 - 14 1933 124
          XM_006233670.2 XP_006233732.1 13 - 14 2037 124
          XM_008761620.1 XP_008759842.1 5 - 6 3793 124
          GenBank mRNA AF107723.1 - 12 - 13 2057 124
          AY136742.1 - 2 - 3 294 124
          BC086535.1 - 3 - 4 493 124

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000097.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpsm1

          Gene Ontology Categories:

          Function(s) Process(es)

          nervous system development
          regulation of G-protein coupled receptor protein signaling pathway
          cell differentiation
          regulation of catalytic activity
          G-protein alpha-subunit binding
          GDP-dissociation inhibitor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007965 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P13393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134949 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5510 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165210 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13393 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007965 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P13393 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007965 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P13393 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P409526 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P668354 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13393 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92834 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27651 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10834418 - -
          Affymetrix Rat Gene 1.0 ST Array 10834425 - -
          Affymetrix Rat Gene 1.0 ST Array 10834431 - -
          Affymetrix Rat Gene 1.0 ST Array 10834416 - -
          Affymetrix Rat Gene 1.0 ST Array 10834422 - -
          Affymetrix Rat Gene 1.0 ST Array 10834419 - -
          Affymetrix Rat Gene 1.0 ST Array 10834414 - -
          Affymetrix Rat Gene 1.0 ST Array 10834412 - -
          Affymetrix Rat Gene 1.0 ST Array 10834420 - -
          Affymetrix Rat Gene 1.0 ST Array 10834423 - -
          Affymetrix Rat Gene 1.0 ST Array 10834424 - -
          Affymetrix Rat Gene 1.0 ST Array 10834415 - -
          Affymetrix Rat Gene 1.0 ST Array 10834421 - -
          Affymetrix Rat Gene 1.0 ST Array 10834429 - -
          Affymetrix Rat Gene 1.0 ST Array 10834413 - -
          Affymetrix Rat Gene 1.0 ST Array 10834430 - -
          Affymetrix Rat Gene 1.0 ST Array 10834428 - -
          Affymetrix Rat Gene 1.0 ST Array 10834426 - -
          Affymetrix Rat Gene 1.0 ST Array 10834417 - -
          Affymetrix Rat Gene 1.0 ST Array 10834427 - -
          Affymetrix Rat Gene 1.1 ST Array 10834411 - -
          Rat Genome 230 2.0 Array 1370590_PM_at - -
          Rat Genome 230 2.0 Array 1372383_at - -
          Rat Genome 230 2.0 Array 1372383_PM_at - -
          Rat Genome 230 2.0 Array 1370590_at - -
          Affymetrix HT Rat Focus Array 1372383_at - -
          Affymetrix HT Rat Focus Array 1370590_PM_at - -
          Affymetrix HT Rat Focus Array 1370590_at - -
          Affymetrix HT Rat Focus Array 1372383_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300000892.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000895.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000894.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000898.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000890.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000893.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000897.rn.1 TC0300000097.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300000896.rn.1 TC0300000097.rn.1 RTA TC
          Rat Expression Set 230 1370590_at - -
          Rat Expression Set 230 1370590_PM_at - -
          Rat Expression Set 230 1372383_at - -
          Rat Expression Set 230 1372383_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10834411 - -
          Rat Genome U34 Set rc_AI229036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22146399 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357506 - -
          RatRef-12 Expression BeadChip ILMN_49324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline