Search
Search
| Gene Symbol: Klhl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145671.1 | NP_663704.1 | 10 - 11 | 1580 | 67 | |
| XM_006239521.2 | XP_006239583.1 | 10 - 11 | 1637 | 67 | ||
| XM_006239526.2 | XP_006239588.1 | 10 - 11 | 1775 | 67 | ||
| XM_008764336.1 | XP_008762558.1 | 10 - 11 | 1637 | 67 | ||
| GenBank mRNA | AF505655.1 | - | 10 - 11 | 1580 | 67 | |
Set Membership: |
|