Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00686175_m1
          See other TNFSF10 GE Assays ›
          Gene Symbol:
          Tnfsf10
          Gene Name:
          tumor necrosis factor (ligand) superfamily, member 10
          Gene Aliases:
          Trail
          Chromosome Location:
          Chr.2: 113008324 - 113024535 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00686175_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfsf10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145681.1 NP_663714.1 4 - 5 435 62
          GenBank mRNA AY115578.1 - 4 - 5 435 62
          EF030546.1 - 4 - 5 426 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200000891.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfsf10

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          male gonad development
          response to insulin
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of extrinsic apoptotic signaling pathway
          molecular_function
          cytokine activity
          tumor necrosis factor receptor binding
          tumor necrosis factor receptor superfamily binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P733671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868102 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122445 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171995 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733671 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P733671 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P733671 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P733671 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73470 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10814530 - -
          Affymetrix Rat Gene 1.0 ST Array 10814533 - -
          Affymetrix Rat Gene 1.0 ST Array 10814529 - -
          Affymetrix Rat Gene 1.0 ST Array 10814531 - -
          Affymetrix Rat Gene 1.0 ST Array 10814532 - -
          Affymetrix Rat Gene 1.1 ST Array 10814528 - -
          Affymetrix Rat Clariom S Assay PSR0200005398.rn.1 TC0200000891.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10814528 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21982054 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362036 - -
          RatRef-12 Expression BeadChip ILMN_52679 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline