Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00686338_m1
          See other PPP1R32 GE Assays ›
          Gene Symbol:
          Ppp1r32
          Gene Name:
          protein phosphatase 1, regulatory subunit 32
          Gene Aliases:
          Iiig9
          Chromosome Location:
          Chr.1: 226518407 - 226526982 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00686338_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145786.1 NP_665729.1 11 - 12 1158 88
          XM_006231016.2 XP_006231078.1 10 - 11 1050 88
          XM_008760216.1 XP_008758438.1 11 - 12 1264 88
          GenBank mRNA AY032664.1 - 10 - 11 1022 88
          AY032665.1 - 11 - 12 1158 88
          BC081718.1 - 11 - 12 1149 7112534 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007253.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r32

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator phosphatase modulator

          Gene Ontology Categories:

          Process(es)

          phosphatase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P205528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P205528 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P205528 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P205528 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P205528 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10728709 - -
          Affymetrix Rat Gene 1.0 ST Array 10728706 - -
          Affymetrix Rat Gene 1.0 ST Array 10728712 - -
          Affymetrix Rat Gene 1.0 ST Array 10728717 - -
          Affymetrix Rat Gene 1.0 ST Array 10728704 - -
          Affymetrix Rat Gene 1.0 ST Array 10728710 - -
          Affymetrix Rat Gene 1.0 ST Array 10728711 - -
          Affymetrix Rat Gene 1.0 ST Array 10728716 - -
          Affymetrix Rat Gene 1.0 ST Array 10728705 - -
          Affymetrix Rat Gene 1.0 ST Array 10728708 - -
          Affymetrix Rat Gene 1.0 ST Array 10728715 - -
          Affymetrix Rat Gene 1.0 ST Array 10728713 - -
          Affymetrix Rat Gene 1.0 ST Array 10728707 - -
          Affymetrix Rat Gene 1.0 ST Array 10728714 - -
          Affymetrix Rat Gene 1.1 ST Array 10728703 - -
          Rat Genome 230 2.0 Array 1370486_a_at - -
          Rat Genome 230 2.0 Array 1370486_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370486_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370486_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100058600.rn.1 TC0100007253.rn.1 RTA TC
          Rat Expression Set 230 1370486_PM_a_at - -
          Rat Expression Set 230 1370486_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10728703 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21979300 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353844 - -
          RatRef-12 Expression BeadChip ILMN_52168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline