Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00687044_m1
          See other NALCN GE Assays ›
          Gene Symbol:
          Nalcn
          Gene Name:
          sodium leak channel, non-selective
          Gene Aliases:
          Vgcnl1
          Chromosome Location:
          Chr.15: 109734092 - 110046729 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00687044_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nalcn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153630.1 NP_705894.1 5080 88
          GenBank mRNA AF078779.1 - 5080 88
          AY555273.1 - 5101 88
          EF492985.1 - 41 - 42 4966 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500002450.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel 3' Most Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nalcn

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sperm motility
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          calcium ion import
          calcium ion transmembrane transport
          membrane depolarization during action potential
          cation transmembrane transport
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          cation channel activity
          sodium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018437 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P018430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126406 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329616 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 259531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 390590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757403 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702494 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 232560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185771 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52335 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 103406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41158 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140054 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159895 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87799 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 48754 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76193 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110630 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166104 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52965 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7216 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40352 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169402 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1264 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119919 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45888 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38826 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81690 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 170436 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32184 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34065 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146704 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018430 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018437 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P018430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018430 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P018437 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1020482 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702137951 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93789 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78826 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72697 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3097 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14702 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34442 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80273 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18377 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52890 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71330 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98730 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 65495 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51288 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30569 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1077 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31085 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48198 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20527 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37392 - -
          Agilent Rat cDNA Microarray (G4105A) 702137951 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10786081 - -
          Affymetrix Rat Gene 1.0 ST Array 10782183 - -
          Affymetrix Rat Gene 1.0 ST Array 10782186 - -
          Affymetrix Rat Gene 1.0 ST Array 10786079 - -
          Affymetrix Rat Gene 1.0 ST Array 10786066 - -
          Affymetrix Rat Gene 1.0 ST Array 10786094 - -
          Affymetrix Rat Gene 1.0 ST Array 10786095 - -
          Affymetrix Rat Gene 1.0 ST Array 10786087 - -
          Affymetrix Rat Gene 1.0 ST Array 10786076 - -
          Affymetrix Rat Gene 1.0 ST Array 10786065 - -
          Affymetrix Rat Gene 1.0 ST Array 10782182 - -
          Affymetrix Rat Gene 1.0 ST Array 10786096 - -
          Affymetrix Rat Gene 1.0 ST Array 10786075 - -
          Affymetrix Rat Gene 1.0 ST Array 10786086 - -
          Affymetrix Rat Gene 1.0 ST Array 10786084 - -
          Affymetrix Rat Gene 1.0 ST Array 10786091 - -
          Affymetrix Rat Gene 1.0 ST Array 10786070 - -
          Affymetrix Rat Gene 1.0 ST Array 10786097 - -
          Affymetrix Rat Gene 1.0 ST Array 10782184 - -
          Affymetrix Rat Gene 1.0 ST Array 10786067 - -
          Affymetrix Rat Gene 1.0 ST Array 10786092 - -
          Affymetrix Rat Gene 1.0 ST Array 10786078 - -
          Affymetrix Rat Gene 1.0 ST Array 10786077 - -
          Affymetrix Rat Gene 1.0 ST Array 10786090 - -
          Affymetrix Rat Gene 1.0 ST Array 10786063 - -
          Affymetrix Rat Gene 1.0 ST Array 10786072 - -
          Affymetrix Rat Gene 1.0 ST Array 10786099 - -
          Affymetrix Rat Gene 1.0 ST Array 10786068 - -
          Affymetrix Rat Gene 1.0 ST Array 10786098 - -
          Affymetrix Rat Gene 1.0 ST Array 10786074 - -
          Affymetrix Rat Gene 1.0 ST Array 10786082 - -
          Affymetrix Rat Gene 1.0 ST Array 10786089 - -
          Affymetrix Rat Gene 1.0 ST Array 10786083 - -
          Affymetrix Rat Gene 1.0 ST Array 10786088 - -
          Affymetrix Rat Gene 1.0 ST Array 10786064 - -
          Affymetrix Rat Gene 1.0 ST Array 10786069 - -
          Affymetrix Rat Gene 1.0 ST Array 10786080 - -
          Affymetrix Rat Gene 1.0 ST Array 10786093 - -
          Affymetrix Rat Gene 1.0 ST Array 10786085 - -
          Affymetrix Rat Gene 1.0 ST Array 10782185 - -
          Affymetrix Rat Gene 1.0 ST Array 10786071 - -
          Affymetrix Rat Gene 1.1 ST Array 10782181 - -
          Affymetrix Rat Gene 1.1 ST Array 10786062 - -
          Rat Genome 230 2.0 Array 1370555_at - -
          Rat Genome 230 2.0 Array 1370555_PM_at - -
          Affymetrix HT Rat Focus Array 1370555_PM_at - -
          Affymetrix HT Rat Focus Array 1370555_at - -
          Affymetrix Rat Clariom S Assay PSR1500016424.rn.1 TC1500002450.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500016425.rn.1 TC1500002450.rn.1 RTA TC
          Rat Expression Set 230 1370555_PM_at - -
          Rat Expression Set 230 1370555_at - -
          Affymetrix Rat Gene 1.0 ST Array 10786062 - -
          Affymetrix Rat Gene 1.0 ST Array 10782181 - -
          Rat Genome U34 Set AF078779_at - -
          Rat Genome U34 Set AF078779_g_at - -
          Rat Genome U34 Set AF078779_g_at - -
          Rat Genome U34 Set AF078779_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21711408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline