Search Thermo Fisher Scientific
Gene Symbol: Tcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012670.1 | NP_036802.1 | 1 - 2 | 142 | 112 | |
GenBank mRNA | D90345.1 | - | 1 - 2 | 142 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |