Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00688897_m1
          See other PLAGL1 GE Assays ›
          Gene Symbol:
          Plagl1
          Gene Name:
          pleiomorphic adenoma gene-like 1
          Gene Aliases:
          Lot1, Zac1
          Chromosome Location:
          Chr.1: 7219587 - 7270169 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00688897_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plagl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012760.3 NP_036892.1 6 - 7 703 74
          GenBank mRNA U72620.1 - 6 - 7 703 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000081.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plagl1

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          regulation of gene expression
          cell differentiation
          skeletal muscle cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15235 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P147225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886708 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639904 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144121 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51770 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47625 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147225 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15235 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P147225 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15235 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P147225 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39675 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37536 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64347 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69831 - -
          Agilent Rat cDNA Microarray (G4105A) 700177980 - -
          Agilent Rat cDNA Microarray (G4105A) 702125510 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10701805 - -
          Affymetrix Rat Gene 1.0 ST Array 10701807 - -
          Affymetrix Rat Gene 1.0 ST Array 10701809 - -
          Affymetrix Rat Gene 1.0 ST Array 10701812 - -
          Affymetrix Rat Gene 1.0 ST Array 10701804 - -
          Affymetrix Rat Gene 1.0 ST Array 10701806 - -
          Affymetrix Rat Gene 1.0 ST Array 10701803 - -
          Affymetrix Rat Gene 1.0 ST Array 10701808 - -
          Affymetrix Rat Gene 1.0 ST Array 10701813 - -
          Affymetrix Rat Gene 1.0 ST Array 10701810 - -
          Affymetrix Rat Gene 1.1 ST Array 10701802 - -
          Rat Genome 230 2.0 Array 1393782_at - -
          Rat Genome 230 2.0 Array 1393782_PM_at - -
          Rat Genome 230 2.0 Array 1387122_at - -
          Rat Genome 230 2.0 Array 1387122_PM_at - -
          Affymetrix HT Rat Focus Array 1387122_at - -
          Affymetrix HT Rat Focus Array 1387122_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100000359.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000367.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000366.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000358.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000368.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000361.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000365.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000370.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000360.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000362.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000364.rn.1 TC0100000081.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000371.rn.1 TC0100000081.rn.1 RTA TC
          Rat Expression Set 230 1387122_PM_at - -
          Rat Expression Set 230 1387122_at - -
          Affymetrix Rat Gene 1.0 ST Array 10701802 - -
          Rat Genome U34 Set U72620_at - -
          Rat Genome U34 Set rc_AA900750_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21304157 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364222 - -
          RatRef-12 Expression BeadChip ILMN_58363 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline