Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689134_g1
          See other CACNB3 GE Assays ›
          Gene Symbol:
          Cacnb3
          Gene Name:
          calcium channel, voltage-dependent, beta 3 subunit
          Gene Aliases:
          CACH3B
          Chromosome Location:
          Chr.7: 140311140 - 140324902 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689134_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cacnb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012828.2 NP_036960.1 12 - 13 1222 118
          XM_006257329.2 XP_006257391.1 12 - 13 1294 118
          XM_006257330.2 XP_006257392.1 12 - 13 1703 118
          GenBank mRNA M88751.1 - 12 - 13 1226 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001783.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cacnb3

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          chemical synaptic transmission
          neuromuscular junction development
          T cell receptor signaling pathway
          calcium ion transmembrane transport
          regulation of voltage-gated calcium channel activity
          voltage-gated calcium channel activity
          protein binding
          high voltage-gated calcium channel activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P245314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803856 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40867 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P245314 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P245314 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P245314 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P245314 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701747992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25616 - -
          Agilent Rat cDNA Microarray (G4105A) 701747992 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899034 - -
          Affymetrix Rat Gene 1.0 ST Array 10899031 - -
          Affymetrix Rat Gene 1.0 ST Array 10899030 - -
          Affymetrix Rat Gene 1.0 ST Array 10899024 - -
          Affymetrix Rat Gene 1.0 ST Array 10899025 - -
          Affymetrix Rat Gene 1.0 ST Array 10899032 - -
          Affymetrix Rat Gene 1.0 ST Array 10899026 - -
          Affymetrix Rat Gene 1.0 ST Array 10899035 - -
          Affymetrix Rat Gene 1.0 ST Array 10899029 - -
          Affymetrix Rat Gene 1.0 ST Array 10899033 - -
          Affymetrix Rat Gene 1.0 ST Array 10899027 - -
          Affymetrix Rat Gene 1.0 ST Array 10899028 - -
          Affymetrix Rat Gene 1.1 ST Array 10899023 - -
          Rat Genome 230 2.0 Array 1387042_at - -
          Rat Genome 230 2.0 Array 1387042_PM_at - -
          Affymetrix HT Rat Focus Array 1387042_PM_at - -
          Affymetrix HT Rat Focus Array 1387042_at - -
          Affymetrix Rat Clariom S Assay PSR0700013707.rn.1 TC0700001783.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700013706.rn.1 TC0700001783.rn.1 RTA TC
          Rat Expression Set 230 1387042_at - -
          Rat Expression Set 230 1387042_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899023 - -
          Rat Genome U34 Set M88751_at - -
          Rat Genome U34 Set M88751_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21242672 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354834 - -
          RatRef-12 Expression BeadChip ILMN_63172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline