Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689219_m1
          See other LAMP1 GE Assays ›
          Gene Symbol:
          Lamp1
          Gene Name:
          lysosomal-associated membrane protein 1
          Gene Aliases:
          LGP120
          Chromosome Location:
          Chr.16: 81689576 - 81706561 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689219_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012857.2 NP_036989.1 6 - 7 951 87
          GenBank mRNA FQ221686.1 - 1 - 2 81 87
          M34959.1 - 6 - 7 951 87
          X14765.1 - 5 - 6 787 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600002017.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lamp1

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          granzyme-mediated apoptotic signaling pathway
          positive regulation of natural killer cell degranulation
          positive regulation of natural killer cell mediated cytotoxicity
          autophagic cell death
          protein stabilization
          establishment of protein localization to organelle
          Golgi to lysosome transport
          regulation of organelle transport along microtubule
          molecular_function
          enzyme binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P421650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511329 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 528342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 348372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596716 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116535 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172300 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P421650 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P421650 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P421650 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700250858 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46257 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5571 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81461 - -
          Agilent Rat cDNA Microarray (G4105A) 700250858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10792786 - -
          Affymetrix Rat Gene 1.0 ST Array 10792779 - -
          Affymetrix Rat Gene 1.0 ST Array 10792776 - -
          Affymetrix Rat Gene 1.0 ST Array 10792777 - -
          Affymetrix Rat Gene 1.0 ST Array 10792778 - -
          Affymetrix Rat Gene 1.0 ST Array 10792781 - -
          Affymetrix Rat Gene 1.0 ST Array 10792785 - -
          Affymetrix Rat Gene 1.0 ST Array 10792780 - -
          Affymetrix Rat Gene 1.0 ST Array 10792784 - -
          Affymetrix Rat Gene 1.0 ST Array 10792788 - -
          Affymetrix Rat Gene 1.0 ST Array 10792787 - -
          Affymetrix Rat Gene 1.1 ST Array 10792775 - -
          Rat Genome 230 2.0 Array 1398816_PM_at - -
          Rat Genome 230 2.0 Array 1368504_PM_at - -
          Rat Genome 230 2.0 Array 1368504_at - -
          Rat Genome 230 2.0 Array 1398816_at - -
          Rat Genome 230 2.0 Array 1375629_PM_at - -
          Rat Genome 230 2.0 Array 1375629_at - -
          Affymetrix HT Rat Focus Array 1398816_at - -
          Affymetrix HT Rat Focus Array 1368504_at - -
          Affymetrix HT Rat Focus Array 1398816_PM_at - -
          Affymetrix HT Rat Focus Array 1368504_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600014925.rn.1 TC1600002017.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600014926.rn.1 TC1600002017.rn.1 RTA TC
          Rat Expression Set 230 1368504_at - -
          Rat Expression Set 230 1375629_at - -
          Rat Expression Set 230 1368504_PM_at - -
          Rat Expression Set 230 1375629_PM_at - -
          Rat Expression Set 230 1398816_PM_at - -
          Rat Expression Set 230 1398816_at - -
          Affymetrix Rat Gene 1.0 ST Array 10792775 - -
          Rat Genome U34 Set rc_AI233362_at - -
          Rat Genome U34 Set rc_AA899808_at - -
          Rat Genome U34 Set rc_AA899808_g_at - -
          Rat Genome U34 Set rc_AA850756_at - -
          Rat Genome U34 Set rc_AI175901_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21584452 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357538 - -
          RatRef-12 Expression BeadChip ILMN_63580 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline