Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00689222_m1
          See other LIPE GE Assays ›
          Gene Symbol:
          Lipe
          Gene Name:
          lipase, hormone sensitive
          Gene Aliases:
          HSL
          Chromosome Location:
          Chr.1: 82248031 - 82266727 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00689222_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lipe
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012859.1 NP_036991.1 2 - 3 1603 94
          XM_006228391.2 XP_006228453.1 2 - 3 736 94
          XM_006228393.2 XP_006228455.1 2 - 3 987 94
          XM_006228394.2 XP_006228456.1 2 - 3 2314 94
          XM_008758895.1 XP_008757117.1 2 - 3 720 94
          XM_008758896.1 XP_008757118.1 2 - 3 880 94
          GenBank mRNA U40001.1 - 2 - 3 1603 94
          X51415.1 - 1 - 2 1125 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004994.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lipe

          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription initiation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          female pregnancy
          cholesterol metabolic process
          lipid catabolic process
          triglyceride catabolic process
          response to drug
          long-chain fatty acid catabolic process
          diacylglycerol catabolic process
          triglyceride lipase activity
          protein binding
          hydrolase activity, acting on ester bonds
          protein kinase binding
          hormone-sensitive lipase activity
          rRNA primary transcript binding
          acylglycerol lipase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P023029 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P338784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690959 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31371 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P338784 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023029 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P023029 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P338784 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P023029 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P338784 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P843424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363106 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26141 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3734 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58543 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719835 - -
          Affymetrix Rat Gene 1.0 ST Array 10705185 - -
          Affymetrix Rat Gene 1.0 ST Array 10719836 - -
          Affymetrix Rat Gene 1.0 ST Array 10719832 - -
          Affymetrix Rat Gene 1.0 ST Array 10719841 - -
          Affymetrix Rat Gene 1.0 ST Array 10719831 - -
          Affymetrix Rat Gene 1.0 ST Array 10719830 - -
          Affymetrix Rat Gene 1.0 ST Array 10719837 - -
          Affymetrix Rat Gene 1.0 ST Array 10719839 - -
          Affymetrix Rat Gene 1.0 ST Array 10719833 - -
          Affymetrix Rat Gene 1.0 ST Array 10705186 - -
          Affymetrix Rat Gene 1.0 ST Array 10719834 - -
          Affymetrix Rat Gene 1.0 ST Array 10705187 - -
          Affymetrix Rat Gene 1.0 ST Array 10719838 - -
          Affymetrix Rat Gene 1.1 ST Array 10705184 - -
          Affymetrix Rat Gene 1.1 ST Array 10719829 - -
          Rat Genome 230 2.0 Array 1382465_at - -
          Rat Genome 230 2.0 Array 1392187_at - -
          Rat Genome 230 2.0 Array 1392187_PM_at - -
          Rat Genome 230 2.0 Array 1382465_PM_at - -
          Rat Genome 230 2.0 Array 1387132_at - -
          Rat Genome 230 2.0 Array 1387132_PM_at - -
          Affymetrix HT Rat Focus Array 1387132_PM_at - -
          Affymetrix HT Rat Focus Array 1382465_PM_at - -
          Affymetrix HT Rat Focus Array 1382465_at - -
          Affymetrix HT Rat Focus Array 1387132_at - -
          Affymetrix Rat Clariom S Assay PSR0100039655.rn.1 TC0100004994.rn.1 RTA TC
          Rat Expression Set 230 1387132_at - -
          Rat Expression Set 230 1387132_PM_at - -
          Rat Expression Set 230 1382465_at - -
          Rat Expression Set 230 1382465_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719829 - -
          Rat Genome U34 Set U40001_at - -
          Rat Genome U34 Set X51415cds_s_at - -
          Rat Genome U34 Set rc_AI176967_at - -
          Rat Genome U34 Set rc_AI236589_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21688955 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1352555 - -
          RatRef-12 Expression BeadChip ILMN_66930 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0