Search
Search
| Gene Symbol: Pde4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113328.1 | NP_001106799.1 | 10 - 11 | 1439 | 64 | |
| NM_001113329.1 | NP_001106800.1 | 11 - 12 | 1354 | 64 | ||
| NM_001113332.1 | NP_001106803.1 | 10 - 11 | 1291 | 64 | ||
| NM_001113334.1 | NP_001106805.1 | 10 - 11 | 1128 | 64 | ||
| NM_017032.1 | NP_058728.1 | 10 - 11 | 1046 | 64 | ||
| XM_008760721.1 | XP_008758943.1 | 10 - 11 | 2447 | 64 | ||
| XM_008760722.1 | XP_008758944.1 | 5 - 6 | 8150 | 64 | ||
| XM_008760723.1 | XP_008758945.1 | 6 - 7 | 790 | 64 | ||
| GenBank mRNA | AF031373.1 | - | 10 - 11 | 1439 | 64 | |
| AF536974.1 | - | 5 - 6 | 762 | 64 | ||
| AF536979.1 | - | 11 - 12 | 1354 | 64 | ||
| AY388961.1 | - | 10 - 11 | 1064 | 64 | ||
| EF102484.1 | - | 10 - 11 | 1291 | 64 | ||
| EF121818.1 | - | 10 - 11 | 1128 | 64 | ||
| L27059.1 | - | 7 - 8 | 765 | 64 | ||
| L27060.1 | - | 5 - 6 | 683 | 64 | ||
| U09455.1 | - | 5 - 6 | 932 | 64 | ||
| U09456.1 | - | 6 - 7 | 790 | 64 | ||
| U09457.1 | - | 10 - 11 | 1046 | 64 | ||
Set Membership: |
|