Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00690623_m1
          See other PLA2G16 GE Assays ›
          Gene Symbol:
          Pla2g16
          Gene Name:
          phospholipase A2, group XVI
          Gene Aliases:
          HRSL3, Hrasls3, Hrev107
          Chromosome Location:
          Chr.1: 222844238 - 222877180 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00690623_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017060.2 NP_058756.2 3 - 4 195 100
          XM_006230687.2 XP_006230749.1 2 - 3 1064 100
          GenBank mRNA AB298805.1 - 1 - 2 114 100
          CB613004.1 - 3 - 4 195 100
          FQ224126.1 - 3 - 4 255 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100003136.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g16

          Gene Ontology Categories:

          Function(s) Process(es)

          phospholipid metabolic process
          phospholipid biosynthetic process
          lipid catabolic process
          negative regulation of cell cycle
          molecular_function
          phospholipase A2 activity
          phosphatidylcholine 1-acylhydrolase activity
          transferase activity, transferring acyl groups
          phosphatidylserine 1-acylhydrolase activity
          1-acyl-2-lysophosphatidylserine acylhydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P159519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 837411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59857 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100488 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159519 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P159519 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P159519 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139749 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P993027 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79684 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57994 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98948 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43291 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10713539 - -
          Affymetrix Rat Gene 1.0 ST Array 10713540 - -
          Affymetrix Rat Gene 1.0 ST Array 10713541 - -
          Affymetrix Rat Gene 1.1 ST Array 10713538 - -
          Rat Genome 230 2.0 Array 1370202_PM_at - -
          Rat Genome 230 2.0 Array 1370202_at - -
          Affymetrix HT Rat Focus Array 1370202_PM_at - -
          Affymetrix HT Rat Focus Array 1370202_at - -
          Affymetrix Rat Clariom S Assay PSR0100025666.rn.1 TC0100003136.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025659.rn.1 TC0100003136.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025665.rn.1 TC0100003136.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025667.rn.1 TC0100003136.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025668.rn.1 TC0100003136.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100025661.rn.1 TC0100003136.rn.1 RTA TC
          Rat Expression Set 230 1370202_PM_at - -
          Rat Expression Set 230 1370202_at - -
          Affymetrix Rat Gene 1.0 ST Array 10713538 - -
          Rat Genome U34 Set X76453_at - -
          Rat Genome U34 Set rc_AA875291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21549672 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376347 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0