Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00691054_m1
          See other CD37 GE Assays ›
          Gene Symbol:
          Cd37
          Gene Name:
          CD37 molecule
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 101230470 - 101236050 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00691054_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017124.1 NP_058820.1 6 - 7 738 75
          XM_006228981.2 XP_006229043.1 7 - 8 931 75
          XM_008759326.1 XP_008757548.1 6 - 7 777 75
          GenBank mRNA BC059144.1 - 6 - 7 779 6887407 75
          FQ230385.1 - 6 - 7 756 75
          X53517.1 - 6 - 7 738 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005384.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd37

          Gene Ontology Categories:

          Function(s)

          positive regulation of immunoglobulin production
          regulation of humoral immune response
          negative regulation of cell proliferation
          negative regulation of myeloid dendritic cell activation
          defense response to protozoan
          regulation of defense response to virus

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P508379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586585 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508379 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P508379 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P508379 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508379 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700189630 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702136626 - -
          Agilent Rat cDNA Microarray (G4105A) 702136626 - -
          Agilent Rat cDNA Microarray (G4105A) 700189630 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10721736 - -
          Affymetrix Rat Gene 1.0 ST Array 10721737 - -
          Affymetrix Rat Gene 1.0 ST Array 10721730 - -
          Affymetrix Rat Gene 1.0 ST Array 10721729 - -
          Affymetrix Rat Gene 1.0 ST Array 10721732 - -
          Affymetrix Rat Gene 1.0 ST Array 10721733 - -
          Affymetrix Rat Gene 1.0 ST Array 10721734 - -
          Affymetrix Rat Gene 1.0 ST Array 10721731 - -
          Affymetrix Rat Gene 1.0 ST Array 10721735 - -
          Affymetrix Rat Gene 1.1 ST Array 10721728 - -
          Rat Genome 230 2.0 Array 1368555_at - -
          Rat Genome 230 2.0 Array 1368555_PM_at - -
          Affymetrix HT Rat Focus Array 1368555_at - -
          Affymetrix HT Rat Focus Array 1368555_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100043819.rn.1 TC0100005384.rn.1 RTA TC
          Rat Expression Set 230 1368555_at - -
          Rat Expression Set 230 1368555_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10721728 - -
          Rat Genome U34 Set X53517_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21544974 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360038 - -
          RatRef-12 Expression BeadChip ILMN_62890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline