Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00692576_m1
          See other SLC12A1 GE Assays ›
          Gene Symbol:
          Slc12a1
          Gene Name:
          solute carrier family 12 (sodium/potassium/chloride transporter), member 1
          Gene Aliases:
          Nkcc2
          Chromosome Location:
          Chr.3: 117421530 - 117498372 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00692576_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc12a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270617.1 NP_001257546.1 3 - 4 757 63
          NM_001270618.1 NP_001257547.1 3 - 4 757 63
          NR_073053.1 3 - 4 799 63
          XM_008762140.1 XP_008760362.1 3 - 4 829 63
          XM_008762141.1 XP_008760363.1 3 - 4 828 63
          XM_008762142.1 XP_008760364.1 3 - 4 829 63
          XM_008762143.1 XP_008760365.1 3 - 4 829 63
          XM_008762144.1 XP_008760366.1 3 - 4 829 63
          GenBank mRNA BC087017.1 - 3 - 4 799 7109809 63
          EF577032.1 - 2 - 3 741 63
          FM059245.1 - 2 - 3 389 63
          JN579708.1 - 3 - 4 757 63
          U10096.1 - 3 - 4 757 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001623.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          potassium ion transport
          sodium ion transport
          chloride transport
          excretion
          diterpenoid metabolic process
          protein insertion into membrane from inner side
          ion transmembrane transport
          renal rubidium ion absorption
          chemical homeostasis
          renal sodium ion absorption
          cation transmembrane transport
          chloride transmembrane transport
          sodium:potassium:chloride symporter activity
          cation:chloride symporter activity
          rubidium ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615018 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237235 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4932 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 102776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27094 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164182 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115744 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149002 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85028 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105013 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007973 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007973 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007973 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P471442 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701827585 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84129 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15054 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89615 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11879 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85126 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31852 - -
          Agilent Rat cDNA Microarray (G4105A) 701827585 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839387 - -
          Affymetrix Rat Gene 1.0 ST Array 10839386 - -
          Affymetrix Rat Gene 1.0 ST Array 10839369 - -
          Affymetrix Rat Gene 1.0 ST Array 10839394 - -
          Affymetrix Rat Gene 1.0 ST Array 10839384 - -
          Affymetrix Rat Gene 1.0 ST Array 10839376 - -
          Affymetrix Rat Gene 1.0 ST Array 10839397 - -
          Affymetrix Rat Gene 1.0 ST Array 10839399 - -
          Affymetrix Rat Gene 1.0 ST Array 10839377 - -
          Affymetrix Rat Gene 1.0 ST Array 10839378 - -
          Affymetrix Rat Gene 1.0 ST Array 10839393 - -
          Affymetrix Rat Gene 1.0 ST Array 10839368 - -
          Affymetrix Rat Gene 1.0 ST Array 10839374 - -
          Affymetrix Rat Gene 1.0 ST Array 10839379 - -
          Affymetrix Rat Gene 1.0 ST Array 10839382 - -
          Affymetrix Rat Gene 1.0 ST Array 10839385 - -
          Affymetrix Rat Gene 1.0 ST Array 10839372 - -
          Affymetrix Rat Gene 1.0 ST Array 10839383 - -
          Affymetrix Rat Gene 1.0 ST Array 10839390 - -
          Affymetrix Rat Gene 1.0 ST Array 10839395 - -
          Affymetrix Rat Gene 1.0 ST Array 10839380 - -
          Affymetrix Rat Gene 1.0 ST Array 10839396 - -
          Affymetrix Rat Gene 1.0 ST Array 10839392 - -
          Affymetrix Rat Gene 1.0 ST Array 10839371 - -
          Affymetrix Rat Gene 1.0 ST Array 10839381 - -
          Affymetrix Rat Gene 1.0 ST Array 10839370 - -
          Affymetrix Rat Gene 1.0 ST Array 10839398 - -
          Affymetrix Rat Gene 1.0 ST Array 10839391 - -
          Affymetrix Rat Gene 1.0 ST Array 10839389 - -
          Affymetrix Rat Gene 1.1 ST Array 10839367 - -
          Rat Genome 230 2.0 Array 1368548_at - -
          Rat Genome 230 2.0 Array 1368548_PM_at - -
          Affymetrix HT Rat Focus Array 1368548_PM_at - -
          Affymetrix HT Rat Focus Array 1368548_at - -
          Affymetrix Rat Clariom S Assay PSR0300011360.rn.1 TC0300001623.rn.1 RTA TC
          Rat Expression Set 230 1368548_at - -
          Rat Expression Set 230 1368548_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839367 - -
          Rat Genome U34 Set U10096_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376627 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0