Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270889.1 | NP_001257818.1 | 4 - 5 | 893 | 58 | |
NM_019144.2 | NP_062017.2 | 4 - 5 | 835 | 58 | ||
XM_006242691.2 | XP_006242753.1 | 6 - 7 | 1541 | 58 | ||
XM_006242692.2 | XP_006242754.1 | 5 - 6 | 1486 | 58 | ||
XM_006242693.2 | XP_006242755.1 | 4 - 5 | 1352 | 58 | ||
XM_006242694.2 | XP_006242756.1 | 4 - 5 | 789 | 58 | ||
GenBank mRNA | BC078847.1 | - | 4 - 5 | 893 | 7110955 | 58 |
BM386942.1 | - | 1 - 2 | 58 | |||
M76110.1 | - | 4 - 5 | 833 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |