Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00694410_g1
          See other STMN3 GE Assays ›
          Gene Symbol:
          Stmn3
          Gene Name:
          stathmin-like 3
          Gene Aliases:
          Sclip
          Chromosome Location:
          Chr.3: 176808016 - 176816108 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00694410_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stmn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024346.1 NP_077322.1 5 - 6 697 106
          GenBank mRNA AY004290.1 - 5 - 6 697 106
          FQ212062.1 - 4 - 5 558 106
          FQ213831.1 - 4 - 5 552 106
          FQ214184.1 - 4 - 5 554 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300005050.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stmn3

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule depolymerization
          regulation of microtubule polymerization or depolymerization
          cytoplasmic microtubule organization
          neuron projection development
          negative regulation of Rac protein signal transduction
          regulation of cytoskeleton organization
          protein binding
          protein C-terminus binding
          tubulin binding
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130207 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059775 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059775 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059775 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1042234 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700526077 - -
          Agilent Rat cDNA Microarray (G4105A) 700526077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10852536 - -
          Affymetrix Rat Gene 1.0 ST Array 10852534 - -
          Affymetrix Rat Gene 1.0 ST Array 10852532 - -
          Affymetrix Rat Gene 1.0 ST Array 10852537 - -
          Affymetrix Rat Gene 1.0 ST Array 10852533 - -
          Affymetrix Rat Gene 1.0 ST Array 10852538 - -
          Affymetrix Rat Gene 1.1 ST Array 10852531 - -
          Rat Genome 230 2.0 Array 1368157_PM_at - -
          Rat Genome 230 2.0 Array 1368157_at - -
          Affymetrix HT Rat Focus Array 1368157_at - -
          Affymetrix HT Rat Focus Array 1368157_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300038406.rn.1 TC0300005050.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300038407.rn.1 TC0300005050.rn.1 RTA TC
          Rat Expression Set 230 1368157_at - -
          Rat Expression Set 230 1368157_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10852531 - -
          Rat Genome U34 Set rc_AA899649_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21149196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355500 - -
          RatRef-12 Expression BeadChip ILMN_55064 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline