Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00694669_m1
          See other ALDH3A1 GE Assays ›
          Gene Symbol:
          Aldh3a1
          Gene Name:
          aldehyde dehydrogenase 3 family, member A1
          Gene Aliases:
          AHDC, Ahd-c, Aldh, Aldh3
          Chromosome Location:
          Chr.10: 47490168 - 47499855 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00694669_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh3a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031972.1 NP_114178.1 7 - 8 1119 101
          GenBank mRNA BC070924.1 - 7 - 8 1152 7096634 101
          J03637.1 - 7 - 8 1119 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000000778.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh3a1

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          cellular aldehyde metabolic process
          aging
          response to nutrient
          positive regulation of cell proliferation
          response to organic cyclic compound
          response to drug
          response to glucocorticoid
          response to cAMP
          oxidation-reduction process
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD) activity
          aldehyde dehydrogenase [NAD(P)+] activity
          alcohol dehydrogenase (NADP+) activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P067654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P067654 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P067654 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12705 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700300759 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78159 - -
          Agilent Rat cDNA Microarray (G4105A) 700300759 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10734239 - -
          Affymetrix Rat Gene 1.0 ST Array 10734234 - -
          Affymetrix Rat Gene 1.0 ST Array 10734230 - -
          Affymetrix Rat Gene 1.0 ST Array 10734232 - -
          Affymetrix Rat Gene 1.0 ST Array 10734235 - -
          Affymetrix Rat Gene 1.0 ST Array 10734237 - -
          Affymetrix Rat Gene 1.0 ST Array 10734236 - -
          Affymetrix Rat Gene 1.0 ST Array 10734231 - -
          Affymetrix Rat Gene 1.0 ST Array 10734238 - -
          Affymetrix Rat Gene 1.0 ST Array 10734229 - -
          Affymetrix Rat Gene 1.0 ST Array 10734233 - -
          Affymetrix Rat Gene 1.1 ST Array 10734228 - -
          Rat Genome 230 2.0 Array 1368130_PM_at - -
          Rat Genome 230 2.0 Array 1368130_at - -
          Affymetrix HT Rat Focus Array 1368130_at - -
          Affymetrix HT Rat Focus Array 1368130_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000006563.rn.1 TC1000000778.rn.1 RTA TC
          Rat Expression Set 230 1368130_at - -
          Rat Expression Set 230 1368130_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10734228 - -
          Rat Genome U34 Set J03637_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20859138 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357303 - -
          RatRef-12 Expression BeadChip ILMN_53723 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline