Search
Search

Ptprf| Gene Symbol: Ptprf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019249.1 | NP_062122.1 | 13 - 14 | 3028 | 70 | |
| XM_006238677.2 | XP_006238739.1 | 16 - 17 | 4071 | 70 | ||
| XM_006238678.2 | XP_006238740.1 | 15 - 16 | 4044 | 70 | ||
| XM_006238680.2 | XP_006238742.1 | 15 - 16 | 4044 | 70 | ||
| XM_006238681.2 | XP_006238743.1 | 14 - 15 | 3738 | 70 | ||
| GenBank mRNA | L11586.1 | - | 13 - 14 | 3028 | 70 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176154 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11993 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11993 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11993 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11993 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700330008 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700372884 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702170807 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84562 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46862 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5786 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27894 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700330008 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702170807 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700372884 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10879159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879151 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879147 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879156 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879168 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879148 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879169 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879144 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879140 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879142 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879155 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879143 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879145 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879150 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879165 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879149 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10879134 | - | - |
| Rat Genome 230 2.0 Array | 1368036_at | - | - |
| Rat Genome 230 2.0 Array | 1371086_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368035_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1368036_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371086_at | - | - |
| Rat Genome 230 2.0 Array | 1368035_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368035_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368036_at | - | - |
| Affymetrix HT Rat Focus Array | 1371086_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368036_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368035_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1371086_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500025482.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025500.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025475.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025472.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025508.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025466.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025501.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025471.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025473.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025469.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025474.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025476.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025506.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025503.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025487.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025483.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025484.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025493.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025481.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025468.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025489.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025507.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025495.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025490.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025465.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025497.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025486.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025488.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025491.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025498.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025478.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025467.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025485.rn.1 | TC0500003452.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500025505.rn.1 | TC0500003452.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368036_PM_at | - | - |
| Rat Expression Set 230 | 1368035_PM_a_at | - | - |
| Rat Expression Set 230 | 1368035_a_at | - | - |
| Rat Expression Set 230 | 1371086_at | - | - |
| Rat Expression Set 230 | 1368036_at | - | - |
| Rat Expression Set 230 | 1371086_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10879134 | - | - |
| Rat Genome U34 Set | M60103_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376592 | - | - |