Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00695953_m1
          See other YWHAB GE Assays ›
          Gene Symbol:
          Ywhab
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 160391108 - 160399931 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00695953_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ywhab
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019377.1 NP_062250.1 4 - 5 746 105
          GenBank mRNA BC076502.1 - 4 - 5 740 7100393 105
          D17446.1 - 4 - 5 741 105
          FQ214109.1 - 4 - 5 782 105
          FQ229978.1 - 4 - 5 751 105
          S55223.1 - 4 - 5 741 105
          S83440.1 - 4 - 5 746 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002268.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ywhab

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          negative regulation of protein dephosphorylation
          positive regulation of catalytic activity
          negative regulation of transcription, DNA-templated
          cytoplasmic sequestering of protein
          protein heterooligomerization
          transcription corepressor activity
          protein binding
          protein C-terminus binding
          enzyme binding
          protein domain specific binding
          protein complex binding
          histone deacetylase binding
          phosphoserine binding
          phosphoprotein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P195306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950195 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141051 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P195306 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P195306 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P195306 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255254 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P195306 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700518016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67187 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31173 - -
          Agilent Rat cDNA Microarray (G4105A) 700518016 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842058 - -
          Affymetrix Rat Gene 1.0 ST Array 10842054 - -
          Affymetrix Rat Gene 1.0 ST Array 10842055 - -
          Affymetrix Rat Gene 1.0 ST Array 10842053 - -
          Affymetrix Rat Gene 1.0 ST Array 10842056 - -
          Affymetrix Rat Gene 1.0 ST Array 10842057 - -
          Affymetrix Rat Gene 1.1 ST Array 10842052 - -
          Rat Genome 230 2.0 Array 1386999_PM_at - -
          Rat Genome 230 2.0 Array 1386999_at - -
          Rat Genome 230 2.0 Array 1398800_PM_at - -
          Rat Genome 230 2.0 Array 1398800_at - -
          Affymetrix HT Rat Focus Array 1398800_PM_at - -
          Affymetrix HT Rat Focus Array 1398800_at - -
          Affymetrix HT Rat Focus Array 1386999_at - -
          Affymetrix HT Rat Focus Array 1386999_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300016938.rn.1 TC0300002268.rn.1 RTA TC
          Rat Expression Set 230 1398800_PM_at - -
          Rat Expression Set 230 1386999_PM_at - -
          Rat Expression Set 230 1398800_at - -
          Rat Expression Set 230 1386999_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842052 - -
          Rat Genome U34 Set S55223_s_at - -
          Rat Genome U34 Set rc_AA924084_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22311031 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350344 - -
          RatRef-12 Expression BeadChip ILMN_65200 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline