Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00696671_m1
          See other CDC42 GE Assays ›
          Gene Symbol:
          Cdc42
          Gene Name:
          cell division cycle 42
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 155690267 - 155728384 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00696671_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cdc42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_171994.4 NP_741991.3 2 - 3 178 75
          XM_008764286.1 XP_008762508.1 2 - 3 683 75
          XM_008764287.1 XP_008762509.1 2 - 3 258 75
          GenBank mRNA AF491841.1 - 1 - 2 171 75
          BC060535.1 - 2 - 3 178 6920523 75
          FQ213007.1 - 2 - 3 177 75
          FQ225045.1 - 2 - 3 177 75
          FQ225541.1 - 2 - 3 290 75
          FQ225902.1 - 2 - 3 177 75
          FQ225973.1 - 2 - 3 290 75
          FQ226943.1 - 2 - 3 173 75
          FQ227167.1 - 2 - 3 177 75
          FQ227499.1 - 2 - 3 177 75
          FQ227894.1 - 2 - 3 177 75
          FQ228149.1 - 2 - 3 290 75
          FQ230725.1 - 2 - 3 186 75
          FQ231482.1 - 2 - 3 186 75
          FQ231846.1 - 2 - 3 290 75
          FQ232179.1 - 2 - 3 177 75
          FQ232295.1 - 2 - 3 186 75
          FQ233586.1 - 2 - 3 188 75
          FQ234217.1 - 2 - 3 177 75
          FQ234639.1 - 2 - 3 186 75
          FQ234830.1 - 2 - 3 186 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500003881.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cdc42

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          sprouting angiogenesis
          cardiac conduction system development
          keratinocyte development
          endocytosis
          actin filament organization
          Golgi organization
          regulation of mitotic nuclear division
          nuclear migration
          small GTPase mediated signal transduction
          metabolic process
          response to glucose
          positive regulation of gene expression
          negative regulation of gene expression
          single organismal cell-cell adhesion
          regulation of exocytosis
          substantia nigra development
          establishment of cell polarity
          actin cytoskeleton organization
          hair follicle morphogenesis
          positive regulation of pseudopodium assembly
          negative regulation of protein complex assembly
          keratinization
          regulation of protein stability
          positive regulation of cytokinesis
          positive regulation of peptidyl-serine phosphorylation
          adherens junction organization
          cellular protein localization
          establishment or maintenance of apical/basal cell polarity
          multicellular organism growth
          dendritic cell migration
          regulation of protein catabolic process
          positive regulation of catalytic activity
          positive regulation of MAPK cascade
          regulation of protein heterodimerization activity
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of DNA replication
          regulation of protein kinase activity
          positive regulation of JNK cascade
          filopodium assembly
          neuron fate determination
          epidermis morphogenesis
          actin filament bundle assembly
          regulation of protein metabolic process
          regulation of filopodium assembly
          nucleus localization
          establishment of Golgi localization
          positive regulation of synapse structural plasticity
          regulation of attachment of spindle microtubules to kinetochore
          heart contraction
          canonical Wnt signaling pathway
          submandibular salivary gland formation
          epithelial-mesenchymal cell signaling
          hair follicle placode formation
          positive regulation of hair follicle cell proliferation
          organelle transport along microtubule
          actin filament branching
          epithelial cell-cell adhesion
          positive regulation of intracellular protein transport
          positive regulation of substrate adhesion-dependent cell spreading
          GTPase activity
          protein binding
          GTP binding
          protein kinase binding
          GTP-dependent protein binding
          mitogen-activated protein kinase kinase kinase binding
          thioesterase binding
          apolipoprotein A-I receptor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P154982 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870043 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820132 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 410565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 16430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151232 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98549 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87833 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154982 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P154982 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133202 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133202 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P154982 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555019 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P555021 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50692 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5785 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82118 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10880778 - -
          Affymetrix Rat Gene 1.0 ST Array 10880783 - -
          Affymetrix Rat Gene 1.0 ST Array 10880781 - -
          Affymetrix Rat Gene 1.0 ST Array 10880784 - -
          Affymetrix Rat Gene 1.0 ST Array 10880780 - -
          Affymetrix Rat Gene 1.0 ST Array 10880782 - -
          Affymetrix Rat Gene 1.1 ST Array 10880777 - -
          Rat Genome 230 2.0 Array 1367475_PM_at - -
          Rat Genome 230 2.0 Array 1370825_PM_a_at - -
          Rat Genome 230 2.0 Array 1367475_at - -
          Rat Genome 230 2.0 Array 1378832_PM_at - -
          Rat Genome 230 2.0 Array 1370825_a_at - -
          Rat Genome 230 2.0 Array 1391127_at - -
          Rat Genome 230 2.0 Array 1378832_at - -
          Rat Genome 230 2.0 Array 1391127_PM_at - -
          Affymetrix HT Rat Focus Array 1367475_PM_at - -
          Affymetrix HT Rat Focus Array 1370825_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370825_a_at - -
          Affymetrix HT Rat Focus Array 1367475_at - -
          Affymetrix Rat Clariom S Assay PSR0500029199.rn.1 TC0500003881.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500029200.rn.1 TC0500003881.rn.1 RTA TC
          Rat Expression Set 230 1367475_at - -
          Rat Expression Set 230 1367475_PM_at - -
          Rat Expression Set 230 1370825_PM_a_at - -
          Rat Expression Set 230 1370825_a_at - -
          Rat Expression Set 230 1391127_at - -
          Rat Expression Set 230 1391127_PM_at - -
          Rat Expression Set 230 1367475_PM_at - -
          Rat Expression Set 230 1367475_at - -
          Affymetrix Rat Gene 1.0 ST Array 10880777 - -
          Rat Genome U34 Set rc_AA925473_at - -
          Rat Genome U34 Set rc_AI227887_at - -
          Rat Genome U34 Set rc_AI176308_at - -
          Rat Genome U34 Set rc_AI227887_g_at - -
          Rat Genome U34 Set rc_AA925473_g_at - -
          Rat Genome U34 Set rc_AI227676_at - -
          Rat Genome U34 Set rc_AA925473_at - -
          Rat Genome U34 Set rc_AI227887_g_at - -
          Rat Genome U34 Set rc_AA925473_g_at - -
          Rat Genome U34 Set rc_AI176308_at - -
          Rat Genome U34 Set rc_AI227887_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371930 - -
          RatRef-12 Expression BeadChip ILMN_66990 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline