Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709358_m1
          See other STX2 GE Assays ›
          Gene Symbol:
          Stx2
          Gene Name:
          syntaxin 2
          Gene Aliases:
          Epim
          Chromosome Location:
          Chr.12: 31324047 - 31360249 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stx2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012748.1 NP_036880.1 8 - 9 680 83
          XM_006249249.2 XP_006249311.1 8 - 9 838 83
          XM_006249250.2 XP_006249312.1 8 - 9 838 83
          XM_006249253.2 XP_006249315.1 9 - 10 2584 83
          XM_008769188.1 XP_008767410.1 9 - 10 3051 83
          GenBank mRNA L20823.1 - 8 - 9 680 83
          L20888.1 - 8 - 9 680 83
          L20889.1 - 8 - 9 680 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000535.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stx2

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          vesicle targeting
          vesicle fusion
          acrosome reaction
          regulation of gene expression
          vesicle-mediated transport
          microvillus assembly
          cell differentiation
          regulation of blood coagulation
          epithelial cell differentiation
          synaptic vesicle fusion to presynaptic active zone membrane
          response to hydroperoxide
          intracellular transport
          vesicle docking
          digestive tract morphogenesis
          protein oligomerization
          regulation of cytoskeleton organization
          membrane fusion
          SNARE binding
          SNAP receptor activity
          protein binding
          protein dimerization activity
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P056376 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499922 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88085 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296981 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85989 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056376 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152009 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152009 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P056376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P056376 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152009 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P229051 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94406 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10758056 - -
          Affymetrix Rat Gene 1.0 ST Array 10758063 - -
          Affymetrix Rat Gene 1.0 ST Array 10758060 - -
          Affymetrix Rat Gene 1.0 ST Array 10758052 - -
          Affymetrix Rat Gene 1.0 ST Array 10758057 - -
          Affymetrix Rat Gene 1.0 ST Array 10758061 - -
          Affymetrix Rat Gene 1.0 ST Array 10758059 - -
          Affymetrix Rat Gene 1.0 ST Array 10758054 - -
          Affymetrix Rat Gene 1.0 ST Array 10758058 - -
          Affymetrix Rat Gene 1.0 ST Array 10758055 - -
          Affymetrix Rat Gene 1.0 ST Array 10758062 - -
          Affymetrix Rat Gene 1.0 ST Array 10758053 - -
          Affymetrix Rat Gene 1.0 ST Array 10758051 - -
          Affymetrix Rat Gene 1.1 ST Array 10758050 - -
          Rat Genome 230 2.0 Array 1370553_at - -
          Rat Genome 230 2.0 Array 1372832_at - -
          Rat Genome 230 2.0 Array 1372832_PM_at - -
          Rat Genome 230 2.0 Array 1370553_PM_at - -
          Affymetrix HT Rat Focus Array 1370553_at - -
          Affymetrix HT Rat Focus Array 1370553_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200004570.rn.1 TC1200000535.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200004572.rn.1 TC1200000535.rn.1 RTA TC
          Rat Expression Set 230 1370553_PM_at - -
          Rat Expression Set 230 1370553_at - -
          Affymetrix Rat Gene 1.0 ST Array 10758050 - -
          Rat Genome U34 Set L20823_at - -
          Rat Genome U34 Set L20823_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21616350 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358230 - -
          RatRef-12 Expression BeadChip ILMN_69120 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline