Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709403_m1
          See other PRKAR2A GE Assays ›
          Gene Symbol:
          Prkar2a
          Gene Name:
          protein kinase, cAMP dependent regulatory, type II alpha
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 117485948 - 117548768 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709403_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkar2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019264.2 NP_062137.1 6 - 7 797 87
          XM_006243712.2 XP_006243774.1 7 - 8 982 87
          XM_006243713.2 XP_006243775.1 7 - 8 796 87
          XM_008766478.1 XP_008764700.1 6 - 7 747 87
          GenBank mRNA AF533978.1 - 6 - 7 801 87
          J02934.1 - 6 - 7 595 87

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkar2a

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorylation
          regulation of protein kinase activity
          protein heterooligomerization
          response to clozapine
          negative regulation of cAMP-dependent protein kinase activity
          cAMP-dependent protein kinase inhibitor activity
          protein binding
          cAMP-dependent protein kinase regulator activity
          kinase activity
          protein domain specific binding
          cAMP binding
          ubiquitin protein ligase binding
          protein kinase A catalytic subunit binding
          small molecule binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P493802 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 407860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86204 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17008 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111127 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156765 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110223 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493802 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P493802 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P493802 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P493802 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32771 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2792 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48407 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10913239 - -
          Affymetrix Rat Gene 1.0 ST Array 10913236 - -
          Affymetrix Rat Gene 1.0 ST Array 10913229 - -
          Affymetrix Rat Gene 1.0 ST Array 10913230 - -
          Affymetrix Rat Gene 1.0 ST Array 10913233 - -
          Affymetrix Rat Gene 1.0 ST Array 10913231 - -
          Affymetrix Rat Gene 1.0 ST Array 10913238 - -
          Affymetrix Rat Gene 1.0 ST Array 10913237 - -
          Affymetrix Rat Gene 1.0 ST Array 10913232 - -
          Affymetrix Rat Gene 1.0 ST Array 10913235 - -
          Affymetrix Rat Gene 1.0 ST Array 10913234 - -
          Affymetrix Rat Gene 1.1 ST Array 10913228 - -
          Rat Genome 230 2.0 Array 1371059_at - -
          Rat Genome 230 2.0 Array 1373400_at - -
          Rat Genome 230 2.0 Array 1395246_at - -
          Rat Genome 230 2.0 Array 1373400_PM_at - -
          Rat Genome 230 2.0 Array 1378415_at - -
          Rat Genome 230 2.0 Array 1378415_PM_at - -
          Rat Genome 230 2.0 Array 1375620_at - -
          Rat Genome 230 2.0 Array 1371059_PM_at - -
          Rat Genome 230 2.0 Array 1375620_PM_at - -
          Rat Genome 230 2.0 Array 1395246_PM_at - -
          Affymetrix HT Rat Focus Array 1373400_at - -
          Affymetrix HT Rat Focus Array 1373400_PM_at - -
          Affymetrix HT Rat Focus Array 1375620_PM_at - -
          Affymetrix HT Rat Focus Array 1371059_PM_at - -
          Affymetrix HT Rat Focus Array 1371059_at - -
          Affymetrix HT Rat Focus Array 1375620_at - -
          Rat Expression Set 230 1373400_PM_at - -
          Rat Expression Set 230 1371059_PM_at - -
          Rat Expression Set 230 1373400_at - -
          Rat Expression Set 230 1375620_at - -
          Rat Expression Set 230 1375620_PM_at - -
          Rat Expression Set 230 1371059_at - -
          Affymetrix Rat Gene 1.0 ST Array 10913228 - -
          Rat Genome U34 Set rc_AI059291_at - -
          Rat Genome U34 Set rc_AA925274_at - -
          Rat Genome U34 Set rc_AI059291_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22250355 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357333 - -
          RatRef-12 Expression BeadChip ILMN_62995 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline