Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709499_m1
          See other PER3 GE Assays ›
          Gene Symbol:
          Per3
          Gene Name:
          period circadian clock 3
          Gene Aliases:
          period3, rper3
          Chromosome Location:
          Chr.5: 168088126 - 168123482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Per3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023978.2 NP_076468.2 5 - 6 643 72
          XM_006239500.2 XP_006239562.1 5 - 6 1095 72
          XM_006239501.2 XP_006239563.1 5 - 6 1095 72
          XM_006239502.2 XP_006239564.1 5 - 6 1095 72
          XM_006239503.2 XP_006239565.1 5 - 6 1094 72
          XM_006239504.2 XP_006239566.1 5 - 6 1095 72
          XM_006239505.2 XP_006239567.1 5 - 6 1094 72
          GenBank mRNA AB092512.1 - 5 - 6 643 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500004116.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Per3

          Panther Classification:

          Molecular Function -

          transcription cofactor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          signal transduction
          circadian rhythm
          circadian regulation of gene expression
          regulation of circadian sleep/wake cycle, sleep
          signal transducer activity
          kinase binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P127337 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119405 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P119407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78218 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324794 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 698549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 260860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 193721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 755650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642383 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60502 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51497 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127337 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119407 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P127337 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119405 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119407 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119407 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119405 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P127337 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P196198 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16143 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P407318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40412 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10881867 - -
          Affymetrix Rat Gene 1.0 ST Array 10881871 - -
          Affymetrix Rat Gene 1.0 ST Array 10881882 - -
          Affymetrix Rat Gene 1.0 ST Array 10881870 - -
          Affymetrix Rat Gene 1.0 ST Array 10881881 - -
          Affymetrix Rat Gene 1.0 ST Array 10881868 - -
          Affymetrix Rat Gene 1.0 ST Array 10881866 - -
          Affymetrix Rat Gene 1.0 ST Array 10881877 - -
          Affymetrix Rat Gene 1.0 ST Array 10881883 - -
          Affymetrix Rat Gene 1.0 ST Array 10881874 - -
          Affymetrix Rat Gene 1.0 ST Array 10881884 - -
          Affymetrix Rat Gene 1.0 ST Array 10881878 - -
          Affymetrix Rat Gene 1.0 ST Array 10881862 - -
          Affymetrix Rat Gene 1.0 ST Array 10881864 - -
          Affymetrix Rat Gene 1.0 ST Array 10881869 - -
          Affymetrix Rat Gene 1.0 ST Array 10881872 - -
          Affymetrix Rat Gene 1.0 ST Array 10881880 - -
          Affymetrix Rat Gene 1.0 ST Array 10881873 - -
          Affymetrix Rat Gene 1.0 ST Array 10881863 - -
          Affymetrix Rat Gene 1.0 ST Array 10881879 - -
          Affymetrix Rat Gene 1.0 ST Array 10881865 - -
          Affymetrix Rat Gene 1.0 ST Array 10881876 - -
          Affymetrix Rat Gene 1.0 ST Array 10881875 - -
          Affymetrix Rat Gene 1.1 ST Array 10881861 - -
          Rat Genome 230 2.0 Array 1388239_at - -
          Rat Genome 230 2.0 Array 1378745_at - -
          Rat Genome 230 2.0 Array 1388239_PM_at - -
          Rat Genome 230 2.0 Array 1378745_PM_at - -
          Affymetrix HT Rat Focus Array 1388239_at - -
          Affymetrix HT Rat Focus Array 1378745_PM_at - -
          Affymetrix HT Rat Focus Array 1388239_PM_at - -
          Affymetrix HT Rat Focus Array 1378745_at - -
          Affymetrix Rat Clariom S Assay PSR0500031450.rn.1 TC0500004116.rn.1 RTA TC
          Rat Expression Set 230 1388239_PM_at - -
          Rat Expression Set 230 1388239_at - -
          Rat Expression Set 230 1378745_at - -
          Rat Expression Set 230 1378745_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10881861 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21609434 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371382 - -
          RatRef-12 Expression BeadChip ILMN_49735 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline