Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709664_m1
          See other DNMT1 GE Assays ›
          Gene Symbol:
          Dnmt1
          Gene Name:
          DNA (cytosine-5-)-methyltransferase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 21922515 - 21968495 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053354.3 NP_445806.3 5 - 6 592 62
          GenBank mRNA AB012214.1 - 5 - 6 576 62
          D64060.1 - 5 - 6 464 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800002054.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnmt1

          Panther Classification:

          Molecular Function -

          DNA metabolism protein DNA methyltransferase nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          DNA methylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          brain development
          aging
          response to heat
          response to toxic substance
          response to organic substance
          response to ionizing radiation
          maintenance of DNA methylation
          response to lead ion
          DNA methylation on cytosine within a CG sequence
          regulation of gene expression
          positive regulation of gene expression
          response to activity
          gene silencing
          chromatin modification
          neuron differentiation
          response to caffeine
          response to nutrient levels
          response to estradiol
          response to lipopolysaccharide
          DNA methylation on cytosine
          response to vitamin A
          response to testosterone
          cellular response to platelet-derived growth factor stimulus
          response to antidepressant
          regulation of cell proliferation
          response to drug
          DNA hypermethylation
          response to ethanol
          negative regulation of transcription, DNA-templated
          S-adenosylhomocysteine metabolic process
          S-adenosylmethioninamine metabolic process
          S-adenosylmethionine metabolic process
          positive regulation of histone H3-K4 methylation
          negative regulation of histone H3-K9 methylation
          cellular response to amino acid stimulus
          cellular response to lead ion
          cellular response to transforming growth factor beta stimulus
          C-5 methylation of cytosine
          cellular response to nerve growth factor stimulus
          DNA binding
          chromatin binding
          double-stranded DNA binding
          RNA binding
          DNA (cytosine-5-)-methyltransferase activity
          protein binding
          transcription factor binding
          methyltransferase activity
          zinc ion binding
          methyl-CpG binding
          S-adenosylmethionine-dependent methyltransferase activity
          DNA-methyltransferase activity
          protein domain specific binding
          estrogen receptor binding
          histone deacetylase binding
          unmethylated CpG binding
          DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P052680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574987 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454216 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738453 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75256 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24634 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93439 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69529 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7855 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052680 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P052680 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P052680 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P388853 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P125681 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700883582 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702569949 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74210 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88710 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32962 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95892 - -
          Agilent Rat cDNA Microarray (G4105A) 702569949 - -
          Agilent Rat cDNA Microarray (G4105A) 700883582 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10915456 - -
          Affymetrix Rat Gene 1.0 ST Array 10915471 - -
          Affymetrix Rat Gene 1.0 ST Array 10915442 - -
          Affymetrix Rat Gene 1.0 ST Array 10915458 - -
          Affymetrix Rat Gene 1.0 ST Array 10915443 - -
          Affymetrix Rat Gene 1.0 ST Array 10915474 - -
          Affymetrix Rat Gene 1.0 ST Array 10915445 - -
          Affymetrix Rat Gene 1.0 ST Array 10915466 - -
          Affymetrix Rat Gene 1.0 ST Array 10915451 - -
          Affymetrix Rat Gene 1.0 ST Array 10915441 - -
          Affymetrix Rat Gene 1.0 ST Array 10915453 - -
          Affymetrix Rat Gene 1.0 ST Array 10915438 - -
          Affymetrix Rat Gene 1.0 ST Array 10915461 - -
          Affymetrix Rat Gene 1.0 ST Array 10915460 - -
          Affymetrix Rat Gene 1.0 ST Array 10915439 - -
          Affymetrix Rat Gene 1.0 ST Array 10915469 - -
          Affymetrix Rat Gene 1.0 ST Array 10915440 - -
          Affymetrix Rat Gene 1.0 ST Array 10915465 - -
          Affymetrix Rat Gene 1.0 ST Array 10915448 - -
          Affymetrix Rat Gene 1.0 ST Array 10915467 - -
          Affymetrix Rat Gene 1.0 ST Array 10915454 - -
          Affymetrix Rat Gene 1.0 ST Array 10915444 - -
          Affymetrix Rat Gene 1.0 ST Array 10915464 - -
          Affymetrix Rat Gene 1.0 ST Array 10915473 - -
          Affymetrix Rat Gene 1.0 ST Array 10915455 - -
          Affymetrix Rat Gene 1.0 ST Array 10915450 - -
          Affymetrix Rat Gene 1.0 ST Array 10915459 - -
          Affymetrix Rat Gene 1.0 ST Array 10915449 - -
          Affymetrix Rat Gene 1.0 ST Array 10915463 - -
          Affymetrix Rat Gene 1.0 ST Array 10915452 - -
          Affymetrix Rat Gene 1.0 ST Array 10915470 - -
          Affymetrix Rat Gene 1.0 ST Array 10915475 - -
          Affymetrix Rat Gene 1.0 ST Array 10915457 - -
          Affymetrix Rat Gene 1.0 ST Array 10915476 - -
          Affymetrix Rat Gene 1.0 ST Array 10915472 - -
          Affymetrix Rat Gene 1.0 ST Array 10915446 - -
          Affymetrix Rat Gene 1.0 ST Array 10915468 - -
          Affymetrix Rat Gene 1.1 ST Array 10915437 - -
          Rat Genome 230 2.0 Array 1373772_at - -
          Rat Genome 230 2.0 Array 1373772_PM_at - -
          Affymetrix HT Rat Focus Array 1373772_PM_at - -
          Affymetrix HT Rat Focus Array 1373772_at - -
          Affymetrix Rat Clariom S Assay PSR0800016466.rn.1 TC0800002054.rn.1 RTA TC
          Rat Expression Set 230 1373772_PM_at - -
          Rat Expression Set 230 1373772_at - -
          Affymetrix Rat Gene 1.0 ST Array 10915437 - -
          Rat Genome U34 Set AB012214_s_at - -
          Rat Genome U34 Set D64060_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22220177 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350922 - -
          RatRef-12 Expression BeadChip ILMN_50839 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline