Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709709_m1
          See other SLC26A3 GE Assays ›
          Gene Symbol:
          Slc26a3
          Gene Name:
          solute carrier family 26 (anion exchanger), member 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 50725085 - 50766306 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709709_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc26a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053755.2 NP_446207.1 11 - 12 1476 73
          GenBank mRNA AF314820.1 - 8 - 9 1000 73
          AF337809.1 - 11 - 12 1481 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600000567.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc26a3

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          anion transport
          bicarbonate transport
          sperm capacitation
          intracellular pH elevation
          membrane hyperpolarization
          cellular response to cAMP
          sulfate transmembrane transport
          chloride transmembrane transport
          secondary active sulfate transmembrane transporter activity
          bicarbonate transmembrane transporter activity
          chloride transmembrane transporter activity
          anion:anion antiporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P012711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 421908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873521 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238068 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40424 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17524 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 30547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012709 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P012709 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P012709 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P597626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37063 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16114 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10884135 - -
          Affymetrix Rat Gene 1.0 ST Array 10884134 - -
          Affymetrix Rat Gene 1.0 ST Array 10884128 - -
          Affymetrix Rat Gene 1.0 ST Array 10884140 - -
          Affymetrix Rat Gene 1.0 ST Array 10884138 - -
          Affymetrix Rat Gene 1.0 ST Array 10884120 - -
          Affymetrix Rat Gene 1.0 ST Array 10884130 - -
          Affymetrix Rat Gene 1.0 ST Array 10884122 - -
          Affymetrix Rat Gene 1.0 ST Array 10884136 - -
          Affymetrix Rat Gene 1.0 ST Array 10884129 - -
          Affymetrix Rat Gene 1.0 ST Array 10884124 - -
          Affymetrix Rat Gene 1.0 ST Array 10884132 - -
          Affymetrix Rat Gene 1.0 ST Array 10884126 - -
          Affymetrix Rat Gene 1.0 ST Array 10884139 - -
          Affymetrix Rat Gene 1.0 ST Array 10884123 - -
          Affymetrix Rat Gene 1.0 ST Array 10884133 - -
          Affymetrix Rat Gene 1.0 ST Array 10884137 - -
          Affymetrix Rat Gene 1.0 ST Array 10884121 - -
          Affymetrix Rat Gene 1.0 ST Array 10884127 - -
          Affymetrix Rat Gene 1.0 ST Array 10884119 - -
          Affymetrix Rat Gene 1.0 ST Array 10884125 - -
          Affymetrix Rat Gene 1.0 ST Array 10884131 - -
          Affymetrix Rat Gene 1.1 ST Array 10884118 - -
          Rat Genome 230 2.0 Array 1388051_PM_at - -
          Rat Genome 230 2.0 Array 1388051_at - -
          Affymetrix HT Rat Focus Array 1388051_PM_at - -
          Affymetrix HT Rat Focus Array 1388051_at - -
          Affymetrix Rat Clariom S Assay PSR0600004079.rn.1 TC0600000567.rn.1 RTA TC
          Rat Expression Set 230 1388051_PM_at - -
          Rat Expression Set 230 1388051_at - -
          Affymetrix Rat Gene 1.0 ST Array 10884118 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20906352 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366727 - -
          RatRef-12 Expression BeadChip ILMN_51781 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline