Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709852_m1
          See other CBLB GE Assays ›
          Gene Symbol:
          Cblb
          Gene Name:
          Cbl proto-oncogene B, E3 ubiquitin protein ligase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 51037383 - 51202777 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709852_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cblb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133601.1 NP_598285.1 6 - 7 888 64
          XM_006248258.2 XP_006248320.1 6 - 7 962 64
          XM_006248259.2 XP_006248321.1 6 - 7 1045 64
          XM_006248260.2 XP_006248322.1 6 - 7 961 64
          XM_008768626.1 XP_008766848.1 5 - 6 863 64
          XM_008768627.1 XP_008766849.1 6 - 7 1158 64
          GenBank mRNA AB071283.1 - 6 - 7 888 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001487.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cblb

          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell anergy
          immune response
          signal transduction
          cell surface receptor signaling pathway
          response to gravity
          response to hormone
          protein ubiquitination
          regulation of signaling
          intracellular signal transduction
          T cell activation
          positive regulation of protein catabolic process
          negative regulation of alpha-beta T cell proliferation
          negative regulation of T cell receptor signaling pathway
          phosphotyrosine binding
          ubiquitin-protein transferase activity
          signal transducer activity
          calcium ion binding
          zinc ion binding
          ligase activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P198539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25710 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 584511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 920401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535891 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 367225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 822499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 254939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915272 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 7387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342635 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64345 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145003 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18390 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 66277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 41060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145316 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13694 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152198 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141808 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171261 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115866 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110354 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5020 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P198539 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P198539 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P198539 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P198539 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702132045 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70530 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 50726 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3163 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70670 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76173 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15787 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96665 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11957 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20910 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82402 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96868 - -
          Agilent Rat cDNA Microarray (G4105A) 702132045 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753749 - -
          Affymetrix Rat Gene 1.0 ST Array 10753753 - -
          Affymetrix Rat Gene 1.0 ST Array 10753762 - -
          Affymetrix Rat Gene 1.0 ST Array 10753761 - -
          Affymetrix Rat Gene 1.0 ST Array 10753765 - -
          Affymetrix Rat Gene 1.0 ST Array 10753748 - -
          Affymetrix Rat Gene 1.0 ST Array 10753754 - -
          Affymetrix Rat Gene 1.0 ST Array 10753757 - -
          Affymetrix Rat Gene 1.0 ST Array 10753764 - -
          Affymetrix Rat Gene 1.0 ST Array 10753759 - -
          Affymetrix Rat Gene 1.0 ST Array 10753768 - -
          Affymetrix Rat Gene 1.0 ST Array 10753767 - -
          Affymetrix Rat Gene 1.0 ST Array 10753755 - -
          Affymetrix Rat Gene 1.0 ST Array 10753763 - -
          Affymetrix Rat Gene 1.0 ST Array 10753766 - -
          Affymetrix Rat Gene 1.0 ST Array 10753756 - -
          Affymetrix Rat Gene 1.0 ST Array 10753758 - -
          Affymetrix Rat Gene 1.0 ST Array 10753750 - -
          Affymetrix Rat Gene 1.0 ST Array 10753760 - -
          Affymetrix Rat Gene 1.0 ST Array 10753751 - -
          Affymetrix Rat Gene 1.1 ST Array 10753747 - -
          Rat Genome 230 2.0 Array 1371027_PM_at - -
          Rat Genome 230 2.0 Array 1371027_at - -
          Affymetrix HT Rat Focus Array 1371027_at - -
          Affymetrix HT Rat Focus Array 1371027_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100009090.rn.1 TC1100001487.rn.1 RTA TC
          Rat Expression Set 230 1371027_at - -
          Rat Expression Set 230 1371027_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753747 - -
          Rat Genome U34 Set rc_AI072631_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21543984 - -
          ABI Rat Genome Survey Microarray 22183058 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351226 - -
          RatRef-12 Expression BeadChip ILMN_63350 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline