Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709933_m1
          See other ARFGAP1 GE Assays ›
          Gene Symbol:
          Arfgap1
          Gene Name:
          ADP-ribosylation factor GTPase activating protein 1
          Gene Aliases:
          Arf GAP1, Arf1gap
          Chromosome Location:
          Chr.3: 176475847 - 176491198 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709933_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arfgap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145090.3 NP_659558.1 8 - 9 912 59
          XM_006235731.2 XP_006235793.1 8 - 9 1012 59
          XM_006235732.1 XP_006235794.1 8 - 9 953 59
          XM_006235733.2 XP_006235795.1 7 - 8 1221 59
          XM_006235734.2 XP_006235796.1 7 - 8 1220 59
          GenBank mRNA BC070895.1 - 8 - 9 893 7099840 59
          DQ167810.1 - 6 - 7 629 59
          DQ167811.1 - 6 - 7 629 59
          FQ222295.1 - 8 - 9 935 59
          FQ232032.1 - 8 - 9 933 59
          FQ233265.1 - 8 - 9 933 59
          U35776.1 - 8 - 9 912 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002587.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arfgap1

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein transport
          vesicle-mediated transport
          regulation of endocytosis
          positive regulation of GTPase activity
          GTPase activator activity
          protein binding
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P058757 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608238 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157664 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80623 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010224 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058757 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P058757 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010224 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P058757 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010224 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13406 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037796 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700912927 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76803 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2892 - -
          Agilent Rat cDNA Microarray (G4105A) 700912927 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10842917 - -
          Affymetrix Rat Gene 1.0 ST Array 10842916 - -
          Affymetrix Rat Gene 1.0 ST Array 10842907 - -
          Affymetrix Rat Gene 1.0 ST Array 10842919 - -
          Affymetrix Rat Gene 1.0 ST Array 10842908 - -
          Affymetrix Rat Gene 1.0 ST Array 10842914 - -
          Affymetrix Rat Gene 1.0 ST Array 10842910 - -
          Affymetrix Rat Gene 1.0 ST Array 10842904 - -
          Affymetrix Rat Gene 1.0 ST Array 10842918 - -
          Affymetrix Rat Gene 1.0 ST Array 10842915 - -
          Affymetrix Rat Gene 1.0 ST Array 10842911 - -
          Affymetrix Rat Gene 1.0 ST Array 10842912 - -
          Affymetrix Rat Gene 1.0 ST Array 10842913 - -
          Affymetrix Rat Gene 1.0 ST Array 10842909 - -
          Affymetrix Rat Gene 1.1 ST Array 10842903 - -
          Rat Genome 230 2.0 Array 1389492_at - -
          Rat Genome 230 2.0 Array 1376123_PM_at - -
          Rat Genome 230 2.0 Array 1384951_PM_at - -
          Rat Genome 230 2.0 Array 1376123_at - -
          Rat Genome 230 2.0 Array 1370364_at - -
          Rat Genome 230 2.0 Array 1384951_at - -
          Rat Genome 230 2.0 Array 1370364_PM_at - -
          Rat Genome 230 2.0 Array 1389492_PM_at - -
          Affymetrix HT Rat Focus Array 1370364_at - -
          Affymetrix HT Rat Focus Array 1376123_at - -
          Affymetrix HT Rat Focus Array 1376123_PM_at - -
          Affymetrix HT Rat Focus Array 1370364_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300019051.rn.1 TC0300002587.rn.1 RTA TC
          Rat Expression Set 230 1389492_at - -
          Rat Expression Set 230 1370364_PM_at - -
          Rat Expression Set 230 1376123_PM_at - -
          Rat Expression Set 230 1370364_at - -
          Rat Expression Set 230 1376123_at - -
          Rat Expression Set 230 1389492_PM_at - -
          Rat Expression Set 230 1384951_PM_at - -
          Rat Expression Set 230 1384951_at - -
          Affymetrix Rat Gene 1.0 ST Array 10842903 - -
          Rat Genome U34 Set U35776_at - -
          Rat Genome U34 Set rc_AA818820_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21789423 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370142 - -
          RatRef-12 Expression BeadChip ILMN_49323 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline