Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709937_m1
          See other ST6GAL1 GE Assays ›
          Gene Symbol:
          St6gal1
          Gene Name:
          ST6 beta-galactosamide alpha-2,6-sialyltranferase 1
          Gene Aliases:
          Siat1
          Chromosome Location:
          Chr.11: 80927601 - 80981424 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709937_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: St6gal1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001113344.1 NP_001106815.1 4 - 5 983 89
          NM_147205.2 NP_671738.2 3 - 4 339 89
          XM_006248501.1 XP_006248563.1 5 - 6 1064 89
          XM_008768792.1 XP_008767014.1 2 - 3 1126 89
          GenBank mRNA FQ210708.1 - 3 - 4 771 89
          FQ222565.1 - 5 - 6 1532 89
          FQ224146.1 - 5 - 6 1468 89
          M18769.1 - 4 - 5 984 89
          M83143.1 - 3 - 4 339 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001873.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene St6gal1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          N-acetylneuraminate metabolic process
          protein glycosylation
          protein N-linked glycosylation via asparagine
          sialylation
          beta-galactoside alpha-2,6-sialyltransferase activity
          sialyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P050852 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P414311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 849332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141681 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91830 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166868 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119778 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106650 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050852 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P414311 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P414311 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P050852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P050852 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P414311 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226906 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P414311 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700481069 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 18609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 29800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81756 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41589 - -
          Agilent Rat cDNA Microarray (G4105A) 700481069 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10755096 - -
          Affymetrix Rat Gene 1.0 ST Array 10755106 - -
          Affymetrix Rat Gene 1.0 ST Array 10755107 - -
          Affymetrix Rat Gene 1.0 ST Array 10755095 - -
          Affymetrix Rat Gene 1.0 ST Array 10755104 - -
          Affymetrix Rat Gene 1.0 ST Array 10755099 - -
          Affymetrix Rat Gene 1.0 ST Array 10755108 - -
          Affymetrix Rat Gene 1.0 ST Array 10755098 - -
          Affymetrix Rat Gene 1.0 ST Array 10755105 - -
          Affymetrix Rat Gene 1.0 ST Array 10755101 - -
          Affymetrix Rat Gene 1.0 ST Array 10755103 - -
          Affymetrix Rat Gene 1.0 ST Array 10755097 - -
          Affymetrix Rat Gene 1.0 ST Array 10755109 - -
          Affymetrix Rat Gene 1.0 ST Array 10755110 - -
          Affymetrix Rat Gene 1.0 ST Array 10755100 - -
          Affymetrix Rat Gene 1.0 ST Array 10755102 - -
          Affymetrix Rat Gene 1.0 ST Array 10755111 - -
          Affymetrix Rat Gene 1.1 ST Array 10755094 - -
          Rat Genome 230 2.0 Array 1370714_PM_a_at - -
          Rat Genome 230 2.0 Array 1370907_at - -
          Rat Genome 230 2.0 Array 1370714_a_at - -
          Rat Genome 230 2.0 Array 1370907_PM_at - -
          Affymetrix HT Rat Focus Array 1370714_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370907_PM_at - -
          Affymetrix HT Rat Focus Array 1370714_a_at - -
          Affymetrix HT Rat Focus Array 1370907_at - -
          Affymetrix Rat Clariom S Assay PSR1100011911.rn.1 TC1100001873.rn.1 RTA TC
          Rat Expression Set 230 1370714_PM_a_at - -
          Rat Expression Set 230 1370714_a_at - -
          Rat Expression Set 230 1370907_at - -
          Rat Expression Set 230 1370907_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10755094 - -
          Rat Genome U34 Set M83143_g_at - -
          Rat Genome U34 Set M83143_at - -
          Rat Genome U34 Set rc_AI029138_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21354101 - -
          ABI Rat Genome Survey Microarray 21507854 - -
          ABI Rat Genome Survey Microarray 21796054 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365332 - -
          RatRef-12 Expression BeadChip ILMN_64777 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline