Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00709999_m1
          See other SERPINF1 GE Assays ›
          Gene Symbol:
          Serpinf1
          Gene Name:
          serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1
          Gene Aliases:
          Dmrs91, Pedf
          Chromosome Location:
          Chr.10: 62241750 - 62254145 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00709999_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Serpinf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177927.2 NP_808788.1 6 - 7 827 71
          GenBank mRNA AY216659.1 - 6 - 7 827 71
          BC078686.1 - 6 - 7 815 7110230 71
          FQ218413.1 - 6 - 7 828 71
          FQ219756.1 - 6 - 7 826 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003241.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Serpinf1

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          aging
          short-term memory
          response to acidic pH
          negative regulation of endothelial cell migration
          negative regulation of gene expression
          negative regulation of endopeptidase activity
          positive regulation of neuron projection development
          negative regulation of angiogenesis
          ovulation cycle
          response to arsenic-containing substance
          negative regulation of inflammatory response
          positive regulation of neurogenesis
          retina development in camera-type eye
          negative regulation of epithelial cell proliferation involved in prostate gland development
          cellular response to cobalt ion
          cellular response to retinoic acid
          cellular response to glucose stimulus
          cellular response to dexamethasone stimulus
          negative regulation of neuron death
          serine-type endopeptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P144202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399676 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3120 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144202 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P144202 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P144202 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P144202 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61507 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45857 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744940 - -
          Affymetrix Rat Gene 1.0 ST Array 10744945 - -
          Affymetrix Rat Gene 1.0 ST Array 10744942 - -
          Affymetrix Rat Gene 1.0 ST Array 10744943 - -
          Affymetrix Rat Gene 1.0 ST Array 10744944 - -
          Affymetrix Rat Gene 1.0 ST Array 10744941 - -
          Affymetrix Rat Gene 1.0 ST Array 10744946 - -
          Affymetrix Rat Gene 1.1 ST Array 10744939 - -
          Rat Genome 230 2.0 Array 1381012_PM_at - -
          Rat Genome 230 2.0 Array 1388569_at - -
          Rat Genome 230 2.0 Array 1381012_at - -
          Rat Genome 230 2.0 Array 1388569_PM_at - -
          Affymetrix HT Rat Focus Array 1388569_PM_at - -
          Affymetrix HT Rat Focus Array 1381012_PM_at - -
          Affymetrix HT Rat Focus Array 1381012_at - -
          Affymetrix HT Rat Focus Array 1388569_at - -
          Affymetrix Rat Clariom S Assay PSR1000029106.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029116.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029109.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029107.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029110.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029108.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029111.rn.1 TC1000003241.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000029114.rn.1 TC1000003241.rn.1 RTA TC
          Rat Expression Set 230 1381012_at - -
          Rat Expression Set 230 1388569_at - -
          Rat Expression Set 230 1381012_PM_at - -
          Rat Expression Set 230 1388569_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744939 - -
          Rat Genome U34 Set rc_AA859899_at - -
          Rat Genome U34 Set rc_AI179984_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21799196 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365894 - -
          RatRef-12 Expression BeadChip ILMN_63264 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline