Search
Search

Sdccag8| Gene Symbol: Sdccag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177929.2 | NP_808790.1 | 13 - 14 | 1737 | 67 | |
| XM_006250321.2 | XP_006250383.1 | 13 - 14 | 1882 | 67 | ||
| XM_006250322.2 | XP_006250384.1 | 13 - 14 | 1882 | 67 | ||
| XM_008769788.1 | XP_008768010.1 | 13 - 14 | 1882 | 67 | ||
| XM_008769789.1 | XP_008768011.1 | 13 - 14 | 1882 | 67 | ||
| XM_008769790.1 | XP_008768012.1 | 13 - 14 | 1882 | 67 | ||
| GenBank mRNA | AF352814.1 | - | 13 - 14 | 1737 | 67 | |
| BC086973.1 | - | 13 - 14 | 1733 | 7191324 | 67 | |
Set Membership: |
|