Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710144_m1
          See other MTUS1 GE Assays ›
          Gene Symbol:
          Mtus1
          Gene Name:
          microtubule associated tumor suppressor 1
          Gene Aliases:
          ATIP4, Atip3b, Mtsg1
          Chromosome Location:
          Chr.16: 54240897 - 54386131 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710144_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtus1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178093.2 NP_835194.2 8 - 9 1244 72
          XM_006253180.2 XP_006253242.1 12 - 13 3859 72
          XM_006253181.2 XP_006253243.1 12 - 13 3559 72
          XM_006253182.2 XP_006253244.1 10 - 11 2179 72
          XM_008771280.1 XP_008769502.1 9 - 10 1603 72
          GenBank mRNA AB190507.1 - 8 - 9 1321 72
          AY208915.1 - 8 - 9 1012 72
          AY208916.1 - 7 - 8 730 72
          BC072537.1 - 8 - 9 1244 72
          BC166591.1 - 8 - 9 1293 7938925 72
          EU417559.1 - 11 - 12 3362 72
          GU245886.1 - 10 - 11 1252 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000627.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtus1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of macrophage chemotaxis
          cellular response to peptide hormone stimulus
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097603 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P097598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 793533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 187433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 214031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476176 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 557757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901511 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 619168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163900 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37128 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127611 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097598 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097603 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P097598 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097603 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P097598 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14440 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P556316 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337271 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77503 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10788395 - -
          Affymetrix Rat Gene 1.0 ST Array 10788389 - -
          Affymetrix Rat Gene 1.0 ST Array 10788388 - -
          Affymetrix Rat Gene 1.0 ST Array 10788382 - -
          Affymetrix Rat Gene 1.0 ST Array 10788387 - -
          Affymetrix Rat Gene 1.0 ST Array 10788392 - -
          Affymetrix Rat Gene 1.0 ST Array 10788390 - -
          Affymetrix Rat Gene 1.0 ST Array 10788383 - -
          Affymetrix Rat Gene 1.0 ST Array 10788393 - -
          Affymetrix Rat Gene 1.0 ST Array 10788396 - -
          Affymetrix Rat Gene 1.0 ST Array 10788384 - -
          Affymetrix Rat Gene 1.0 ST Array 10788394 - -
          Affymetrix Rat Gene 1.0 ST Array 10788391 - -
          Affymetrix Rat Gene 1.1 ST Array 10788374 - -
          Rat Genome 230 2.0 Array 1376058_PM_at - -
          Rat Genome 230 2.0 Array 1376058_at - -
          Rat Genome 230 2.0 Array 1384680_PM_at - -
          Rat Genome 230 2.0 Array 1380321_at - -
          Rat Genome 230 2.0 Array 1372457_PM_at - -
          Rat Genome 230 2.0 Array 1380321_PM_at - -
          Rat Genome 230 2.0 Array 1384680_at - -
          Rat Genome 230 2.0 Array 1372457_at - -
          Affymetrix HT Rat Focus Array 1376058_at - -
          Affymetrix HT Rat Focus Array 1372457_at - -
          Affymetrix HT Rat Focus Array 1380321_PM_at - -
          Affymetrix HT Rat Focus Array 1376058_PM_at - -
          Affymetrix HT Rat Focus Array 1380321_at - -
          Affymetrix HT Rat Focus Array 1372457_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600004997.rn.1 TC1600000627.rn.1 RTA TC
          Rat Expression Set 230 1372457_PM_at - -
          Rat Expression Set 230 1376058_at - -
          Rat Expression Set 230 1372457_at - -
          Rat Expression Set 230 1376058_PM_at - -
          Rat Expression Set 230 1380321_PM_at - -
          Rat Expression Set 230 1380321_at - -
          Affymetrix Rat Gene 1.0 ST Array 10788374 - -
          Rat Genome U34 Set rc_AA849422_at - -
          Rat Genome U34 Set rc_AI230939_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21141322 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370195 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-wn6hs:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline