Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710210_m1
          See other PLOD3 GE Assays ›
          Gene Symbol:
          Plod3
          Gene Name:
          procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 22716421 - 22726982 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710210_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plod3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178101.3 NP_835202.2 18 - 19 2352 73
          GenBank mRNA AJ430859.1 - 18 - 19 2091 73
          BC085683.1 - 18 - 19 2132 7110561 73

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plod3

          Panther Classification:

          Molecular Function -

          protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          endothelial cell morphogenesis
          protein localization
          neural tube development
          collagen fibril organization
          cellular response to hormone stimulus
          vasodilation
          epidermis morphogenesis
          oxidation-reduction process
          lung morphogenesis
          basement membrane assembly
          molecular_function
          iron ion binding
          procollagen-lysine 5-dioxygenase activity
          L-ascorbic acid binding
          procollagen glucosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P832915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717848 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9332 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832915 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P832915 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P832915 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P832915 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27646 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757465 - -
          Affymetrix Rat Gene 1.0 ST Array 10757463 - -
          Affymetrix Rat Gene 1.0 ST Array 10757478 - -
          Affymetrix Rat Gene 1.0 ST Array 10757472 - -
          Affymetrix Rat Gene 1.0 ST Array 10757469 - -
          Affymetrix Rat Gene 1.0 ST Array 10757462 - -
          Affymetrix Rat Gene 1.0 ST Array 10757468 - -
          Affymetrix Rat Gene 1.0 ST Array 10757481 - -
          Affymetrix Rat Gene 1.0 ST Array 10757477 - -
          Affymetrix Rat Gene 1.0 ST Array 10757467 - -
          Affymetrix Rat Gene 1.0 ST Array 10757480 - -
          Affymetrix Rat Gene 1.0 ST Array 10757474 - -
          Affymetrix Rat Gene 1.0 ST Array 10757473 - -
          Affymetrix Rat Gene 1.0 ST Array 10757476 - -
          Affymetrix Rat Gene 1.0 ST Array 10757475 - -
          Affymetrix Rat Gene 1.0 ST Array 10757464 - -
          Affymetrix Rat Gene 1.0 ST Array 10757471 - -
          Affymetrix Rat Gene 1.0 ST Array 10757466 - -
          Affymetrix Rat Gene 1.0 ST Array 10757479 - -
          Affymetrix Rat Gene 1.0 ST Array 10757470 - -
          Affymetrix Rat Gene 1.1 ST Array 10757461 - -
          Rat Genome 230 2.0 Array 1372877_at - -
          Rat Genome 230 2.0 Array 1372877_PM_at - -
          Affymetrix HT Rat Focus Array 1372877_at - -
          Affymetrix HT Rat Focus Array 1372877_PM_at - -
          Rat Expression Set 230 1372877_at - -
          Rat Expression Set 230 1372877_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757461 - -
          Rat Genome U34 Set rc_AA892859_at - -
          Rat Genome U34 Set rc_AA892859_g_at - -
          Rat Genome U34 Set rc_AA923945_s_at - -
          Rat Genome U34 Set rc_AI072366_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21773267 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364313 - -
          RatRef-12 Expression BeadChip ILMN_66297 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline