Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00710289_m1
          See other SYTL5 GE Assays ›
          Gene Symbol:
          Sytl5
          Gene Name:
          synaptotagmin-like 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 14017405 - 14109592 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00710289_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sytl5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178333.1 NP_848016.1 6 - 7 831 68
          XM_008773069.1 XP_008771291.1 8 - 9 2161 68
          XM_008773070.1 XP_008771292.1 8 - 9 2152 68
          GenBank mRNA AB098162.1 - 6 - 7 831 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000106.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sytl5

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          exocytosis
          Rab GTPase binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P503240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403712 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403572 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 936821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 535240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 604991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310116 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 48565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 614475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78657 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 4009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7013 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 20689 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19456 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141878 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122976 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119719 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 179949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57465 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503240 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P503240 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P503240 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P503240 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90888 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87053 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54598 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25847 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10936881 - -
          Affymetrix Rat Gene 1.0 ST Array 10936885 - -
          Affymetrix Rat Gene 1.0 ST Array 10936882 - -
          Affymetrix Rat Gene 1.0 ST Array 10936894 - -
          Affymetrix Rat Gene 1.0 ST Array 10936884 - -
          Affymetrix Rat Gene 1.0 ST Array 10936880 - -
          Affymetrix Rat Gene 1.0 ST Array 10936889 - -
          Affymetrix Rat Gene 1.0 ST Array 10936893 - -
          Affymetrix Rat Gene 1.0 ST Array 10936878 - -
          Affymetrix Rat Gene 1.0 ST Array 10936887 - -
          Affymetrix Rat Gene 1.0 ST Array 10936891 - -
          Affymetrix Rat Gene 1.0 ST Array 10936890 - -
          Affymetrix Rat Gene 1.0 ST Array 10936879 - -
          Affymetrix Rat Gene 1.0 ST Array 10936888 - -
          Affymetrix Rat Gene 1.0 ST Array 10936883 - -
          Affymetrix Rat Gene 1.0 ST Array 10936892 - -
          Affymetrix Rat Gene 1.0 ST Array 10936886 - -
          Affymetrix Rat Gene 1.1 ST Array 10936877 - -
          Affymetrix Rat Clariom S Assay PSR0X00000662.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000664.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000665.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000661.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000663.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0X00000666.rn.1 TC0X00000106.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10936877 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22201492 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351920 - -
          RatRef-12 Expression BeadChip ILMN_68941 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline